missing translation for 'onlineSavingsMsg'
Learn More
4-Bromo-2,6-dichloroaniline, 98%, Thermo Scientific Chemicals Product Code.: 11482677

4-Bromo-2,6-dichloroaniline, 98%, Thermo Scientific Chemicals

Product Code. 11482677
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11482677

Brand: Thermo Scientific Alfa Aesar A14884.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 697-88-1
Molecular Formula C6H4BrCl2N
Molecular Weight (g/mol) 240.909
MDL Number MFCD00025162
InChI Key NPQBZKNXJZARBJ-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-bromoaniline, 4-bromo-2,6-dichlorophenyl amine, 4-bromo-2,6-dichlorophenylamine, 4-bromo-2,6-dichloro-benzenamine, benzenamine, 4-bromo-2,6-dichloro, 4-brom-2,6-dichloranilin, pubchem3606, acmc-1ay6t, ksc495a6h, 4-bromo-2,6-dichloro anilineShow MoreShow Less
PubChem CID 69680
IUPAC Name 4-bromo-2,6-dichloroaniline
SMILES C1=C(C=C(C(=C1Cl)N)Cl)Br

Specifications

Melting Point 85°C to 88°C
Quantity 25 g
Beilstein 2089687
Formula Weight 240.92
Percent Purity 98%
Chemical Name or Material 4-Bromo-2,6-dichloroaniline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10159098 Certificate TypeCertificate of Analysis Date05/07/2022 Catalog NumberA14884.22,A14884.14
Lot Number10011423 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14884.22,A14884.14
Lot NumberI2400A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14884.22,A14884.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Bromo-2,6-dichloroaniline, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.