Learn More
4-Bromo-2-methylpyridine, 97%, Thermo Scientific Chemicals
£30.00 - £79.10
Chemical Identifiers
CAS | 22282-99-1 |
---|---|
Molecular Formula | C6H6BrN |
Molecular Weight (g/mol) | 172.03 |
MDL Number | MFCD03788196 |
InChI Key | JFBMFWHEXBLFCR-UHFFFAOYSA-N |
Synonym | 4-bromo-2-picoline, 2-methyl-4-bromopyridine, 4-bromo-2-methyl-pyridine, pyridine, 4-bromo-2-methyl, 4-bromopicoline, zlchem 422, pubchem4035, acmc-209fui, 4-bromo-methyl-pyridine |
PubChem CID | 2762835 |
IUPAC Name | 4-bromo-2-methylpyridine |
SMILES | CC1=CC(Br)=CC=N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15463807
|
Thermo Scientific Alfa Aesar
H27884.MD |
250 mg |
£30.00
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11367509
|
Thermo Scientific Alfa Aesar
H27884.03 |
1 g |
£79.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Bromo-2-methylpyridine is used as a starting material in the preparation of crown-ester-bipyridines and viologens through sodium or nickel reductive coupling, side chain oxidation and esterification. Further, it serves as an important raw material and intermediate in organic synthesis, pharmaceuticals, agrochemicals and in dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromo-2-methylpyridine is used as a starting material in the preparation of crown-ester-bipyridines and viologens through sodium or nickel reductive coupling, side chain oxidation and esterification. Further, it serves as an important raw material and intermediate in organic synthesis, pharmaceuticals, agrochemicals and in dyes.
Solubility
Miscible with dichloromethane. Immiscible with water.
Notes
Light and moisture sensitive. Incompatible with strong oxidizing agents, reducing agents, strong acids and bases. Store in a cool place.
Chemical Identifiers
22282-99-1 | |
172.03 | |
JFBMFWHEXBLFCR-UHFFFAOYSA-N | |
2762835 | |
CC1=CC(Br)=CC=N1 |
C6H6BrN | |
MFCD03788196 | |
4-bromo-2-picoline, 2-methyl-4-bromopyridine, 4-bromo-2-methyl-pyridine, pyridine, 4-bromo-2-methyl, 4-bromopicoline, zlchem 422, pubchem4035, acmc-209fui, 4-bromo-methyl-pyridine | |
4-bromo-2-methylpyridine |
Specifications
22282-99-1 | |
76°C (14 mmHg) | |
Characteristic | |
1.556 | |
4-bromo-2-picoline, 2-methyl-4-bromopyridine, 4-bromo-2-methyl-pyridine, pyridine, 4-bromo-2-methyl, 4-bromopicoline, zlchem 422, pubchem4035, acmc-209fui, 4-bromo-methyl-pyridine | |
JFBMFWHEXBLFCR-UHFFFAOYSA-N | |
4-bromo-2-methylpyridine | |
2762835 | |
97% |
1.45 | |
79°C (174°F) | |
C6H6BrN | |
MFCD03788196 | |
Miscible with dichloromethane. Immiscible with water. | |
CC1=CC(Br)=CC=N1 | |
172.03 | |
172.03 | |
4-Bromo-2-methylpyridine |
Safety and Handling
GHS H Statement
H301-H311-H227-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H319
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only