missing translation for 'onlineSavingsMsg'
Learn More
4-Bromo-N,N-dimethylbenzenesulfonamide, 97%, Thermo Scientific™ Product Code.: 11450050

4-Bromo-N,N-dimethylbenzenesulfonamide, 97%, Thermo Scientific™

Product Code. 11450050
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11450050

Brand: Thermo Scientific Alfa Aesar H50579.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 707-60-8
Molecular Formula C8H10BrNO2S
Molecular Weight (g/mol) 264.137
MDL Number MFCD00017834
InChI Key NQAUNPZZVCXYEJ-UHFFFAOYSA-N
Synonym 4-bromo-n,n-dimethyl-benzenesulfonamide, benzenesulfonamide, 4-bromo-n,n-dimethyl, p-bromo-n,n-dimethylbenzenesulfonamide, 4-bromobenzenedimethylsulfonamide, 4-bromo-n,n-dimethylbenzene-1-sulfonamide, n,n-dimethyl 4-bromobenzenesulfonamide, benzenesulfonamide, p-bromo-n,n-dimethyl, n,n-dimethyl-4-bromobenzenesulfonamide, 4-bromo-n,n-dimethylbenzenesulphonamide, acmc-209og0Show MoreShow Less
PubChem CID 12818
IUPAC Name 4-bromo-N,N-dimethylbenzenesulfonamide
SMILES CN(C)S(=O)(=O)C1=CC=C(C=C1)Br

Specifications

Quantity 1 g
Formula Weight 264.14
Percent Purity 97%
Chemical Name or Material 4-Bromo-N,N-dimethylbenzenesulfonamide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberB02W006 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot NumberJ27Q014 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberL20S055 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Bromo-N,N-dimethylbenzenesulfonamide, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.