missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromomethylbenzenesulfonyl chloride, 95%, Thermo Scientific Chemicals
£17.60 - £182.00
Chemical Identifiers
CAS | 66176-39-4 |
---|---|
Molecular Formula | C7H6BrClO2S |
Molecular Weight (g/mol) | 269.55 |
MDL Number | MFCD00156129 |
InChI Key | QXTQWYZHHMQSQH-UHFFFAOYSA-N |
Synonym | 4-bromomethyl benzenesulfonyl chloride, 4-bromomethyl benzene-1-sulfonyl chloride, 4-bromomethyl benzenesulfonylchloride, alpha-bromo-p-toluenesulphonyl chloride, alpha-bromo-p-toluenesulfonyl chloride, 4-bromomethyl benzenesulphonyl chloride, 4-bromomethyl benzene-1-sulfonylchloride, buttpark 94\04-08, 4-bromomethyl phenyl chlorosulfone, 4-bromomethylene benzenesulfonyl chloride |
PubChem CID | 2734409 |
IUPAC Name | 4-(bromomethyl)benzenesulfonyl chloride |
SMILES | C1=CC(=CC=C1CBr)S(=O)(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10342963
|
Thermo Scientific Acros
360540010 |
1 g | Glass bottle |
£17.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10445292
|
Thermo Scientific Acros
360540050 |
5 g | Glass bottle |
£54.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10116673
|
Thermo Scientific Acros
360540250 |
25 g | Glass bottle |
£182.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
66176-39-4 | |
269.55 | |
QXTQWYZHHMQSQH-UHFFFAOYSA-N | |
2734409 | |
C1=CC(=CC=C1CBr)S(=O)(=O)Cl |
C7H6BrClO2S | |
MFCD00156129 | |
4-bromomethyl benzenesulfonyl chloride, 4-bromomethyl benzene-1-sulfonyl chloride, 4-bromomethyl benzenesulfonylchloride, alpha-bromo-p-toluenesulphonyl chloride, alpha-bromo-p-toluenesulfonyl chloride, 4-bromomethyl benzenesulphonyl chloride, 4-bromomethyl benzene-1-sulfonylchloride, buttpark 94\04-08, 4-bromomethyl phenyl chlorosulfone, 4-bromomethylene benzenesulfonyl chloride | |
4-(bromomethyl)benzenesulfonyl chloride |
Specifications
66176-39-4 | |
100.0 | |
White to Yellow | |
94% min. (Argentometry) | |
BrCH2C6H4SO2Cl | |
4-bromomethyl benzenesulfonyl chloride, 4-bromomethyl benzene-1-sulfonyl chloride, 4-bromomethyl benzenesulfonylchloride, alpha-bromo-p-toluenesulphonyl chloride, alpha-bromo-p-toluenesulfonyl chloride, 4-bromomethyl benzenesulphonyl chloride, 4-bromomethyl benzene-1-sulfonylchloride, buttpark 94\04-08, 4-bromomethyl phenyl chlorosulfone, 4-bromomethylene benzenesulfonyl chloride | |
C1=CC(=CC=C1CBr)S(=O)(=O)Cl | |
269.55 | |
269.55 | |
Crystalline Powder |
94.0 | |
69.0°C to 76.0°C | |
Authentic | |
C7H6BrClO2S | |
MFCD00156129 | |
QXTQWYZHHMQSQH-UHFFFAOYSA-N | |
4-(bromomethyl)benzenesulfonyl chloride | |
2734409 | |
95% | |
4-Bromomethylbenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4-Bromomethylbenzenesulfonyl chloride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only