missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2-methylbenzyl alcohol, 97%, Thermo Scientific™
£58.99 - £167.00
Chemical Identifiers
CAS | 129716-11-6 |
---|---|
Molecular Formula | C8H9ClO |
Molecular Weight (g/mol) | 156.609 |
MDL Number | MFCD06201140 |
InChI Key | AHXBDGJNZJKLNE-UHFFFAOYSA-N |
Synonym | 4-chloro-2-methylphenyl methanol, 4-chloro-2-methylbenzyl alcohol, benzenemethanol, 4-chloro-2-methyl, acmc-20aaxd, 4-chloro-2-methylbenzylalcohol, 4-chloro-2-methyl-phenyl-methanol |
PubChem CID | 2757669 |
IUPAC Name | (4-chloro-2-methylphenyl)methanol |
SMILES | CC1=C(C=CC(=C1)Cl)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11461850
|
Thermo Scientific Alfa Aesar
H27554.03 |
1 g |
£58.99
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11471850
|
Thermo Scientific Alfa Aesar
H27554.06 |
5 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
129716-11-6 | |
156.609 | |
AHXBDGJNZJKLNE-UHFFFAOYSA-N | |
2757669 | |
CC1=C(C=CC(=C1)Cl)CO |
C8H9ClO | |
MFCD06201140 | |
4-chloro-2-methylphenyl methanol, 4-chloro-2-methylbenzyl alcohol, benzenemethanol, 4-chloro-2-methyl, acmc-20aaxd, 4-chloro-2-methylbenzylalcohol, 4-chloro-2-methyl-phenyl-methanol | |
(4-chloro-2-methylphenyl)methanol |
Specifications
129716-11-6 | |
C8H9ClO | |
4-chloro-2-methylphenyl methanol, 4-chloro-2-methylbenzyl alcohol, benzenemethanol, 4-chloro-2-methyl, acmc-20aaxd, 4-chloro-2-methylbenzylalcohol, 4-chloro-2-methyl-phenyl-methanol | |
CC1=C(C=CC(=C1)Cl)CO | |
156.609 | |
156.61 | |
4-Chloro-2-methylbenzyl alcohol |
60°C to 64°C | |
MFCD06201140 | |
AHXBDGJNZJKLNE-UHFFFAOYSA-N | |
(4-chloro-2-methylphenyl)methanol | |
2757669 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 4-Chloro-2-methylbenzyl alcohol
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 4-Chloro-2-methylbenzyl alcohol