missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzoylacetonitrile, 98%, Thermo Scientific Chemicals

£47.00 - £663.00

Chemical Identifiers

CAS 4640-66-8
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.603
MDL Number MFCD00051625
InChI Key JYOUFPNYTOFCSJ-UHFFFAOYSA-N
Synonym 4-chlorobenzoylacetonitrile, 3-4-chlorophenyl-3-oxopropanenitrile, 4-chlorophenacylcyanide, p-chlorophenacyl cyanide, 4-chlorobenzoyl acetonitrile, 3-4-chloro-phenyl-3-oxo-propionitrile, 4-chlorobenzoylacetonitirle, zlchem 1256, pubchem12045, acmc-1akag
PubChem CID 138308
IUPAC Name 3-(4-chlorophenyl)-3-oxopropanenitrile
SMILES C1=CC(=CC=C1C(=O)CC#N)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11459503
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12364.06
5 g
£47.00
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11469503
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12364.14
25 g
£184.00
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11479503
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12364.22
100 g
£663.00
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Pharmaceutical Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pharmaceutical Intermediates.

Solubility
Soluble in Dichloromethane, Ether, Ethyl Acetate and Methanol

Notes
Store at -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

4640-66-8
179.603
JYOUFPNYTOFCSJ-UHFFFAOYSA-N
138308
C1=CC(=CC=C1C(=O)CC#N)Cl
C9H6ClNO
MFCD00051625
4-chlorobenzoylacetonitrile, 3-4-chlorophenyl-3-oxopropanenitrile, 4-chlorophenacylcyanide, p-chlorophenacyl cyanide, 4-chlorobenzoyl acetonitrile, 3-4-chloro-phenyl-3-oxo-propionitrile, 4-chlorobenzoylacetonitirle, zlchem 1256, pubchem12045, acmc-1akag
3-(4-chlorophenyl)-3-oxopropanenitrile

Specifications

4640-66-8
C9H6ClNO
743368
Soluble in Dichloromethane,Ether,Ethyl Acetate and Methanol
C1=CC(=CC=C1C(=O)CC#N)Cl
179.603
179.61
4-Chlorobenzoylacetonitrile
128°C to 132°C
MFCD00051625
4-chlorobenzoylacetonitrile, 3-4-chlorophenyl-3-oxopropanenitrile, 4-chlorophenacylcyanide, p-chlorophenacyl cyanide, 4-chlorobenzoyl acetonitrile, 3-4-chloro-phenyl-3-oxo-propionitrile, 4-chlorobenzoylacetonitirle, zlchem 1256, pubchem12045, acmc-1akag
JYOUFPNYTOFCSJ-UHFFFAOYSA-N
3-(4-chlorophenyl)-3-oxopropanenitrile
138308
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only