Learn More
4-(Di-n-propylsulfamoyl)benzoyl chloride, 96%, Thermo Scientific™
£61.90 - £198.00
Chemical Identifiers
CAS | 29171-72-0 |
---|---|
Molecular Formula | C13H18ClNO3S |
Molecular Weight (g/mol) | 303.80 |
MDL Number | MFCD00121817 |
InChI Key | LCQXGFTXKWEWQC-UHFFFAOYSA-N |
Synonym | 4-dipropylsulfamoyl benzoyl chloride, 4-dipropylamino sulfonyl benzene-1-carbonyl chloride, 4-n,n-dipropylsulfamoyl benzoyl chloride, benzoyl chloride,4-dipropylamino sulfonyl, acmc-1cejw, 4-dipropylsulfamoyl-benzoyl chloride, 4-di-n-propylsulfamoyl benzoyl chloride, 4-dipropylaminosulfonyl benzoyl chloride |
PubChem CID | 2735528 |
IUPAC Name | 4-(dipropylsulfamoyl)benzoyl chloride |
SMILES | CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)C(Cl)=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
29171-72-0 | |
303.80 | |
LCQXGFTXKWEWQC-UHFFFAOYSA-N | |
2735528 | |
CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)C(Cl)=O |
C13H18ClNO3S | |
MFCD00121817 | |
4-dipropylsulfamoyl benzoyl chloride, 4-dipropylamino sulfonyl benzene-1-carbonyl chloride, 4-n,n-dipropylsulfamoyl benzoyl chloride, benzoyl chloride,4-dipropylamino sulfonyl, acmc-1cejw, 4-dipropylsulfamoyl-benzoyl chloride, 4-di-n-propylsulfamoyl benzoyl chloride, 4-dipropylaminosulfonyl benzoyl chloride | |
4-(dipropylsulfamoyl)benzoyl chloride |
Specifications
29171-72-0 | |
C13H18ClNO3S | |
UN3261 | |
4-dipropylsulfamoyl benzoyl chloride, 4-dipropylamino sulfonyl benzene-1-carbonyl chloride, 4-n,n-dipropylsulfamoyl benzoyl chloride, benzoyl chloride,4-dipropylamino sulfonyl, acmc-1cejw, 4-dipropylsulfamoyl-benzoyl chloride, 4-di-n-propylsulfamoyl benzoyl chloride, 4-dipropylaminosulfonyl benzoyl chloride | |
CCCN(CCC)S(=O)(=O)C1=CC=C(C=C1)C(Cl)=O | |
303.80 | |
303.81 | |
4-(Di-n-propylsulfamoyl)benzoyl chloride |
59°C to 61°C | |
MFCD00121817 | |
Moisture sensitive | |
LCQXGFTXKWEWQC-UHFFFAOYSA-N | |
4-(dipropylsulfamoyl)benzoyl chloride | |
2735528 | |
96% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.