missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Dimethoxymethyl-2-(trifluoromethyl)pyrimidine, 97%, Thermo Scientific™
£66.70 - £205.00
Chemical Identifiers
CAS | 878760-47-5 |
---|---|
Molecular Formula | C8H9F3N2O2 |
Molecular Weight (g/mol) | 222.167 |
MDL Number | MFCD05662688 |
InChI Key | CHSZUWDVGYPDPG-UHFFFAOYSA-N |
Synonym | 4-dimethoxymethyl-2-trifluoromethyl pyrimidine, 4-dimethoxymethyl-2-trifluoromethyl-pyrimidine, pyrimidine,4-dimethoxymethyl-2-trifluoromethyl, dimethoxy 2-trifluoromethyl pyrimidin-4-yl methane, pyrimidine, 4-dimethoxymethyl-2-trifluoromethyl |
PubChem CID | 17750125 |
IUPAC Name | 4-(dimethoxymethyl)-2-(trifluoromethyl)pyrimidine |
SMILES | COC(C1=NC(=NC=C1)C(F)(F)F)OC |
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
NotesIncompatible with oxidizing agents.
Chemische Identifikatoren
878760-47-5 | |
222.167 | |
CHSZUWDVGYPDPG-UHFFFAOYSA-N | |
17750125 | |
COC(C1=NC(=NC=C1)C(F)(F)F)OC |
C8H9F3N2O2 | |
MFCD05662688 | |
4-dimethoxymethyl-2-trifluoromethyl pyrimidine, 4-dimethoxymethyl-2-trifluoromethyl-pyrimidine, pyrimidine,4-dimethoxymethyl-2-trifluoromethyl, dimethoxy 2-trifluoromethyl pyrimidin-4-yl methane, pyrimidine, 4-dimethoxymethyl-2-trifluoromethyl | |
4-(dimethoxymethyl)-2-(trifluoromethyl)pyrimidine |
Spezifikation
878760-47-5 | |
MFCD05662688 | |
CHSZUWDVGYPDPG-UHFFFAOYSA-N | |
4-(dimethoxymethyl)-2-(trifluoromethyl)pyrimidine | |
17750125 | |
97% |
C8H9F3N2O2 | |
4-dimethoxymethyl-2-trifluoromethyl pyrimidine, 4-dimethoxymethyl-2-trifluoromethyl-pyrimidine, pyrimidine,4-dimethoxymethyl-2-trifluoromethyl, dimethoxy 2-trifluoromethyl pyrimidin-4-yl methane, pyrimidine, 4-dimethoxymethyl-2-trifluoromethyl | |
COC(C1=NC(=NC=C1)C(F)(F)F)OC | |
222.167 | |
222.17 | |
4-Dimethoxymethyl-2-(trifluoromethyl)pyrimidine |
Sicherheit und Handhabung
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures