missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Dimethylsulfamoyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals
£63.50 - £176.00
Chemical Identifiers
CAS | 486422-59-7 |
---|---|
Molecular Formula | C8H12BNO4S |
Molecular Weight (g/mol) | 229.06 |
MDL Number | MFCD06659849 |
InChI Key | VVUGXBSXXJTVDS-UHFFFAOYSA-N |
Synonym | 4-n,n-dimethylsulfamoyl phenylboronic acid, n,n-dimethyl 4-boronobenzenesulfonamide, 4-n,n-dimethylsulphonamido benzeneboronic acid, 4-dimethylsulfamoyl phenylboronic acid, 4-dimethylsulfamoyl phenyl boronic acid, 4-n,n-dimethylsulfamoyl phenyl boronic acid, 4-borono-n,n-dimethylbenzenesulphonamide, 4-n,n-dimethylsulfonamidophenyl boronic acid, n,n-dimethyl-4-boronobenzenesulfonamide, boronic acid, 4-dimethylamino sulfonyl phenyl |
PubChem CID | 44119328 |
IUPAC Name | [4-(dimethylsulfamoyl)phenyl]boronic acid |
SMILES | CN(C)S(=O)(=O)C1=CC=C(C=C1)B(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15419707
|
Thermo Scientific Alfa Aesar
H52936.MD |
250 mg |
£63.50
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15409707
|
Thermo Scientific Alfa Aesar
H52936.03 |
1 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
486422-59-7 | |
229.06 | |
VVUGXBSXXJTVDS-UHFFFAOYSA-N | |
44119328 | |
CN(C)S(=O)(=O)C1=CC=C(C=C1)B(O)O |
C8H12BNO4S | |
MFCD06659849 | |
4-n,n-dimethylsulfamoyl phenylboronic acid, n,n-dimethyl 4-boronobenzenesulfonamide, 4-n,n-dimethylsulphonamido benzeneboronic acid, 4-dimethylsulfamoyl phenylboronic acid, 4-dimethylsulfamoyl phenyl boronic acid, 4-n,n-dimethylsulfamoyl phenyl boronic acid, 4-borono-n,n-dimethylbenzenesulphonamide, 4-n,n-dimethylsulfonamidophenyl boronic acid, n,n-dimethyl-4-boronobenzenesulfonamide, boronic acid, 4-dimethylamino sulfonyl phenyl | |
[4-(dimethylsulfamoyl)phenyl]boronic acid |
Specifications
486422-59-7 | |
C8H12BNO4S | |
4-n,n-dimethylsulfamoyl phenylboronic acid, n,n-dimethyl 4-boronobenzenesulfonamide, 4-n,n-dimethylsulphonamido benzeneboronic acid, 4-dimethylsulfamoyl phenylboronic acid, 4-dimethylsulfamoyl phenyl boronic acid, 4-n,n-dimethylsulfamoyl phenyl boronic acid, 4-borono-n,n-dimethylbenzenesulphonamide, 4-n,n-dimethylsulfonamidophenyl boronic acid, n,n-dimethyl-4-boronobenzenesulfonamide, boronic acid, 4-dimethylamino sulfonyl phenyl | |
CN(C)S(=O)(=O)C1=CC=C(C=C1)B(O)O | |
229.06 | |
229.06 | |
4-(Dimethylsulfamoyl)benzeneboronic acid |
140°C to 144°C | |
MFCD06659849 | |
VVUGXBSXXJTVDS-UHFFFAOYSA-N | |
[4-(dimethylsulfamoyl)phenyl]boronic acid | |
44119328 | |
95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only