Learn More
4-Fluoro-2-methylphenylboronic acid, 97%, Thermo Scientific™
£33.97 - £519.00
Chemical Identifiers
CAS | 139911-29-8 |
---|---|
MDL Number | MFCD02093072 |
InChI Key | IQMLIVUHMSIOQP-UHFFFAOYSA-N |
Synonym | 2-methyl-4-fluorophenylboronic acid, 4-fluoro-2-methylbenzeneboronic acid, 4-fluoro-2-methylphenyl boronic acid, 4-fluoro-2-methylphenyl boranediol, 4-fluoro-2-methyl-phenyl boronic acid, boronic acid, 4-fluoro-2-methylphenyl, 4-fluoro-2-methylphenylboronicacid, pubchem1852, acmc-209cjp |
PubChem CID | 2734665 |
IUPAC Name | (4-fluoro-2-methylphenyl)boronic acid |
SMILES | B(C1=C(C=C(C=C1)F)C)(O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10611663
|
Thermo Scientific Acros
367940010 |
1 g | Glass bottle | N/A | |||||
10782833
|
Thermo Scientific Acros
367940050 |
5 g | Glass bottle | N/A | |||||
10003393
|
Thermo Scientific Acros
367940250 |
25 g | Glass bottle | N/A | |||||
Description
Chemical Identifiers
139911-29-8 | |
IQMLIVUHMSIOQP-UHFFFAOYSA-N | |
2734665 | |
B(C1=C(C=C(C=C1)F)C)(O)O |
MFCD02093072 | |
2-methyl-4-fluorophenylboronic acid, 4-fluoro-2-methylbenzeneboronic acid, 4-fluoro-2-methylphenyl boronic acid, 4-fluoro-2-methylphenyl boranediol, 4-fluoro-2-methyl-phenyl boronic acid, boronic acid, 4-fluoro-2-methylphenyl, 4-fluoro-2-methylphenylboronicacid, pubchem1852, acmc-209cjp | |
(4-fluoro-2-methylphenyl)boronic acid |
Specifications
139911-29-8 | |
100.0 | |
Authentic | |
C7H8BFO2 | |
2-methyl-4-fluorophenylboronic acid, 4-fluoro-2-methylbenzeneboronic acid, 4-fluoro-2-methylphenyl boronic acid, 4-fluoro-2-methylphenyl boranediol, 4-fluoro-2-methyl-phenyl boronic acid, boronic acid, 4-fluoro-2-methylphenyl, 4-fluoro-2-methylphenylboronicacid, pubchem1852, acmc-209cjp | |
B(C1=C(C=C(C=C1)F)C)(O)O | |
153.947 | |
153.95 | |
Powder |
96.0 | |
White | |
97% | |
MFCD02093072 | |
IQMLIVUHMSIOQP-UHFFFAOYSA-N | |
(4-fluoro-2-methylphenyl)boronic acid | |
2734665 | |
≥96.0% | |
4-Fluoro-2-methylphenylboronic acid |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
- 4-Fluoro-2-methylphenylboronic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only