missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluoro-3-iodobenzoic acid, 97%, Thermo Scientific™
£118.00
Chemical Identifiers
CAS | 403-18-9 |
---|---|
Molecular Formula | C7H4FIO2 |
Molecular Weight (g/mol) | 266.01 |
MDL Number | MFCD03788556 |
InChI Key | IKSOHJRYPQRTLQ-UHFFFAOYSA-N |
Synonym | benzoic acid, 4-fluoro-3-iodo, 3-iodo-4-fluorobenzoic acid, 4-fluoro-3-iodo-benzoic acid, pubchem1390, buttpark 122, buttpark 122\01-04 |
PubChem CID | 10361 |
IUPAC Name | 4-fluoro-3-iodobenzoic acid |
SMILES | C1=CC(=C(C=C1C(=O)O)I)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15466617
|
Thermo Scientific Alfa Aesar
H33762.03 |
1 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
403-18-9 | |
266.01 | |
IKSOHJRYPQRTLQ-UHFFFAOYSA-N | |
10361 | |
C1=CC(=C(C=C1C(=O)O)I)F |
C7H4FIO2 | |
MFCD03788556 | |
benzoic acid, 4-fluoro-3-iodo, 3-iodo-4-fluorobenzoic acid, 4-fluoro-3-iodo-benzoic acid, pubchem1390, buttpark 122, buttpark 122\01-04 | |
4-fluoro-3-iodobenzoic acid |
Specifications
403-18-9 | |
C7H4FIO2 | |
Light sensitive | |
IKSOHJRYPQRTLQ-UHFFFAOYSA-N | |
4-fluoro-3-iodobenzoic acid | |
10361 | |
97% |
173°C to 175°C | |
MFCD03788556 | |
benzoic acid, 4-fluoro-3-iodo, 3-iodo-4-fluorobenzoic acid, 4-fluoro-3-iodo-benzoic acid, pubchem1390, buttpark 122, buttpark 122\01-04 | |
C1=CC(=C(C=C1C(=O)O)I)F | |
266.01 | |
266.01 | |
4-Fluoro-3-iodobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures