missing translation for 'onlineSavingsMsg'
Learn More
4-Hydroxy-3-methoxymandelic acid, 99%, Thermo Scientific Chemicals Product Code.: 10458900

4-Hydroxy-3-methoxymandelic acid, 99%, Thermo Scientific Chemicals

Product Code. 10458900
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10458900

Brand: Thermo Scientific Acros 164900010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-10-7
Molecular Formula C9H9O5
Molecular Weight (g/mol) 197.17
MDL Number MFCD00004235
InChI Key CGQCWMIAEPEHNQ-QMMMGPOBSA-M
Synonym vanillylmandelic acid, vanilmandelic acid, dl-4-hydroxy-3-methoxymandelic acid, 2-hydroxy-2-4-hydroxy-3-methoxyphenyl acetic acid, 4-hydroxy-3-methoxymandelic acid, vanillomandelic acid, +/--vanillylmandelic acid, vma, alpha,4-dihydroxy-3-methoxybenzeneacetic acid, benzeneacetic acid, .alpha.,4-dihydroxy-3-methoxyShow MoreShow Less
PubChem CID 1245
ChEBI CHEBI:20106
SMILES COC1=CC(=CC=C1O)[C@H](O)C([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 131.0°C to 134.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HOC6H3(OCH3)CH(OH)CO2H
Quantity 1 g
Merck Index 15,10121
Solubility Information Solubility in water: freely soluble. Other solubilities: freely soluble in acetone,moderatly soluble in ether and acetonitrile,sparingly soluble in benzene
IUPAC Name 2-hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetic acid
Formula Weight 198.17
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Hydroxy-3-methoxymandelic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0387126 Certificate TypeCertificate of Analysis Date10/08/2022 Catalog Number
Lot NumberA0381415 Certificate TypeCertificate of Analysis Date07/02/2022 Catalog Number
compliance-icons
Product Identifier
  • 4-Hydroxy-3-methoxymandelic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Hydroxy-3-methoxymandelic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.