Learn More
4-(Hydroxymercury)benzoic Acid Sodium Salt, 98%, Thermo Scientific Chemicals
£18.30 - £77.50
Chemical Identifiers
CAS | 138-85-2 |
---|---|
Molecular Formula | C7H5HgNaO3 |
Molecular Weight (g/mol) | 360.69 |
MDL Number | MFCD00002527 |
InChI Key | BERBQUNVJGVZCX-UHFFFAOYSA-M |
Synonym | sodium 4-carboxylatophenyl mercury hydrate, sodium hydrate p-mercuribenzoate |
PubChem CID | 23667696 |
IUPAC Name | sodium;(4-carboxylatophenyl)mercury;hydrate |
SMILES | C1=CC(=CC=C1C(=O)[O-])[Hg].O.[Na+] |
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
138-85-2 | |
360.69 | |
BERBQUNVJGVZCX-UHFFFAOYSA-M | |
23667696 | |
C1=CC(=CC=C1C(=O)[O-])[Hg].O.[Na+] |
C7H5HgNaO3 | |
MFCD00002527 | |
sodium 4-carboxylatophenyl mercury hydrate, sodium hydrate p-mercuribenzoate | |
sodium;(4-carboxylatophenyl)mercury;hydrate |
Specifications
138-85-2 | |
100.0 | |
White | |
97.5% min. (ex Hg) | |
MFCD00002527 | |
sodium 4-carboxylatophenyl mercury hydrate, sodium hydrate p-mercuribenzoate | |
BERBQUNVJGVZCX-UHFFFAOYSA-M | |
sodium;(4-carboxylatophenyl)mercury;hydrate | |
23667696 | |
98% | |
4-(Hydroxymercury)benzoic acid sodium salt |
97.5 | |
>300°C | |
Authentic | |
C7H5HgNaO3 | |
16, 969 | |
Solubility in water: slightly soluble in water | |
C1=CC(=CC=C1C(=O)[O-])[Hg].O.[Na+] | |
360.69 | |
360.69 | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
Fatal if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Fatal in contact with skin.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-340-5
missing translation for 'rtecsNumber' : OV8150000
missing translation for 'tsca' : TSCA
- 4-(Hydroxymercury)benzoic acid sodium salt
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H310-Fatal in contact with skin.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H330-Fatal if inhaled.
- H373-May cause damage to organs.
- H300-Fatal if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P361-Take off immediately all contaminated clothing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.