missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxy-3-(trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals
£55.30 - £175.00
Chemical Identifiers
CAS | 149507-36-8 |
---|---|
Molecular Formula | C8H8BF3O3 |
Molecular Weight (g/mol) | 219.954 |
MDL Number | MFCD07363790 |
InChI Key | BUSMBMGODABSIN-UHFFFAOYSA-N |
Synonym | 4-methoxy-3-trifluoromethyl phenyl boronic acid, 4-methoxy-3-trifluoromethylphenylboronic acid, 3-trifluoromethyl-4-methoxy-phenylboronic acid, 4-methoxy-3-trifluoromethylphenyl boronic acid, 4-methoxy-3-trifluoromethyl benzeneboronic acid, 4-methoxy-3-trifluoromethyl phenylboronic acid, boronic acid, 4-methoxy-3-trifluoromethyl phenyl, acmc-1c5ik |
PubChem CID | 17750052 |
IUPAC Name | [4-methoxy-3-(trifluoromethyl)phenyl]boronic acid |
SMILES | B(C1=CC(=C(C=C1)OC)C(F)(F)F)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15449995
|
Thermo Scientific Alfa Aesar
H52660.MD |
250 mg |
£55.30
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15439995
|
Thermo Scientific Alfa Aesar
H52660.03 |
1 g |
£175.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
149507-36-8 | |
219.954 | |
BUSMBMGODABSIN-UHFFFAOYSA-N | |
17750052 | |
B(C1=CC(=C(C=C1)OC)C(F)(F)F)(O)O |
C8H8BF3O3 | |
MFCD07363790 | |
4-methoxy-3-trifluoromethyl phenyl boronic acid, 4-methoxy-3-trifluoromethylphenylboronic acid, 3-trifluoromethyl-4-methoxy-phenylboronic acid, 4-methoxy-3-trifluoromethylphenyl boronic acid, 4-methoxy-3-trifluoromethyl benzeneboronic acid, 4-methoxy-3-trifluoromethyl phenylboronic acid, boronic acid, 4-methoxy-3-trifluoromethyl phenyl, acmc-1c5ik | |
[4-methoxy-3-(trifluoromethyl)phenyl]boronic acid |
Specifications
149507-36-8 | |
C8H8BF3O3 | |
4-methoxy-3-trifluoromethyl phenyl boronic acid, 4-methoxy-3-trifluoromethylphenylboronic acid, 3-trifluoromethyl-4-methoxy-phenylboronic acid, 4-methoxy-3-trifluoromethylphenyl boronic acid, 4-methoxy-3-trifluoromethyl benzeneboronic acid, 4-methoxy-3-trifluoromethyl phenylboronic acid, boronic acid, 4-methoxy-3-trifluoromethyl phenyl, acmc-1c5ik | |
B(C1=CC(=C(C=C1)OC)C(F)(F)F)(O)O | |
219.954 | |
219.95 | |
4-Methoxy-3-(trifluoromethyl)benzeneboronic acid |
198°C to 202°C | |
MFCD07363790 | |
BUSMBMGODABSIN-UHFFFAOYSA-N | |
[4-methoxy-3-(trifluoromethyl)phenyl]boronic acid | |
17750052 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only