Learn More
4-Methoxyphenol, 99%, Thermo Scientific Chemicals
£15.70 - £1047.00
Chemical Identifiers
CAS | 150-76-5, 123-31-9 |
---|---|
Molecular Formula | C7H8O2 |
Molecular Weight (g/mol) | 124.14 |
MDL Number | MFCD00002332 |
InChI Key | NWVVVBRKAWDGAB-UHFFFAOYSA-N |
Synonym | mequinol, 4-hydroxyanisole, p-hydroxyanisole, p-methoxyphenol, hydroquinone monomethyl ether, phenol, 4-methoxy, leucobasal, mehq, leucodine b, mechinolum |
PubChem CID | 9015 |
ChEBI | CHEBI:69441 |
IUPAC Name | 4-methoxyphenol |
SMILES | COC1=CC=C(O)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10294930
|
Thermo Scientific Acros
126001000 |
100 g |
£15.70
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10479620
|
Thermo Scientific Acros
126005000 |
500 g |
£48.50
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10605562
|
Thermo Scientific Acros
126000010 |
1 kg |
£78.50
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10012923
|
Thermo Scientific Acros
126000051 |
5 kg |
£266.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15234624
|
Thermo Scientific Acros
126000250 |
25 kg |
£1047.00
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
150-76-5, 123-31-9 | |
124.14 | |
NWVVVBRKAWDGAB-UHFFFAOYSA-N | |
9015 | |
4-methoxyphenol |
C7H8O2 | |
MFCD00002332 | |
mequinol, 4-hydroxyanisole, p-hydroxyanisole, p-methoxyphenol, hydroquinone monomethyl ether, phenol, 4-methoxy, leucobasal, mehq, leucodine b, mechinolum | |
CHEBI:69441 | |
COC1=CC=C(O)C=C1 |
Specifications
150-76-5, 123-31-9 | |
100.0 | |
Colorless to Yellow | |
133°C | |
98.5% min. (GC) | |
C7H8O2 | |
06,843 | |
Solubility in water: 40g/L (25°C). Other solubilities: readily soluble in ethyl acetate, 455g/100g ethyl alcohol - 426g/100g acetone (25°C), 70g/100g benzene - 3g/100g carbon tetrachloride, (25°C) | |
COC1=CC=C(O)C=C1 | |
124.14 | |
CHEBI:69441 | |
124.14 | |
Pure | |
4-Methoxyphenol, Stabilized |
98.5 | |
56.0°C to 57.0°C | |
243.0°C | |
Authentic | |
Plastic bottle | |
MFCD00002332 | |
mequinol, 4-hydroxyanisole, p-hydroxyanisole, p-methoxyphenol, hydroquinone monomethyl ether, phenol, 4-methoxy, leucobasal, mehq, leucodine b, mechinolum | |
NWVVVBRKAWDGAB-UHFFFAOYSA-N | |
4-methoxyphenol | |
9015 | |
8.5 mPa.s (65°C) | |
99% | |
Liquid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye irritation.
Suspected of damaging the unborn child.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
If eye i
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 205-769-8
- 4-Methoxyphenol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only