missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanone, ACS Reagent, ≥98.5%, Honeywell™ Riedel-de Haën™

ACS Reagent,≥98.5%

£28.31 - £536.94

Chemical Identifiers

CAS 108-10-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00008938
InChI Key NTIZESTWPVYFNL-UHFFFAOYSA-N
Synonym 4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
PubChem CID 7909
ChEBI CHEBI:82344
IUPAC Name 4-methylpentan-2-one
SMILES CC(C)CC(=O)C
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15645080
View Documents
Honeywell-Riedel-de Haen
360511-500ML
500 mL Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15615080
View Documents
Honeywell-Riedel-de Haen
360511-1L
1 L Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15625080
View Documents
Honeywell-Riedel-de Haen
360511-2.5L
2.5 L Glass Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15635080
View Documents
Honeywell-Riedel-de Haen
360511-20L
20 L Stainless Steel Can This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Specifications

Chemical Identifiers

108-10-1
100.161
NTIZESTWPVYFNL-UHFFFAOYSA-N
7909
4-methylpentan-2-one
C6H12O
MFCD00008938
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
CHEBI:82344
CC(C)CC(=O)C

Specifications

108-10-1
0.801g/mL at 25°C
14°C (57.2°F)
C6H12O
n20/D 1.395
UN1245
4-methyl-2-pentanone, methyl isobutyl ketone, isopropylacetone, isobutyl methyl ketone, hexone, mibk, 2-pentanone, 4-methyl, 2-methyl-4-pentanone, 4-methyl-2-oxopentane, methylisobutylketon
CC(C)CC(=O)C
100.161
CHEBI:82344
≥98.5%
-80°C
118°C
Conforms to structure
(CH3)2CHCH2COCH3
MFCD00008938
605399
NTIZESTWPVYFNL-UHFFFAOYSA-N
4-methylpentan-2-one
7909
100.16g/mol
4-Methyl-2-pentanone
Videos
Safety and Handling

Safety and Handling

P210-P261-P305 + P351 + P338

missing translation for 'dotInformation' : class 3 - PG 2 - Methyl isobutyl ketone

missing translation for 'rtecsNumber' : 203-550-1

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.