missing translation for 'onlineSavingsMsg'
Learn More

4-Morpholinophenylboronic acid, 97%, Thermo Scientific™

£46.00 - £530.00

Chemical Identifiers

CAS 186498-02-2
Molecular Formula C10H14BNO3
Molecular Weight (g/mol) 207.04
MDL Number MFCD03095169
InChI Key WHDIUBHAKZDSJL-UHFFFAOYSA-N
Synonym 4-morpholinophenylboronic acid, 4-morpholinophenyl boronic acid, 4-morpholino phenylboronic acid, 4-morpholin-4-yl-phenylboronic acid, 4-morpholin-4-ylphenyl boronic acid, 4-4-boronophenyl morpholine, 4-morpholino phenyl boronic acid, 4-morpholin-4-yl phenylboronic acid, boronic acid, 4-4-morpholinyl phenyl, boronic acid, b-4-4-morpholinyl phenyl
PubChem CID 2795359
IUPAC Name (4-morpholin-4-ylphenyl)boronic acid
SMILES OB(O)C1=CC=C(C=C1)N1CCOCC1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10279722
View Documents
Thermo Scientific Maybridge
CC17412CB
250 mg Amber Glass Bottle
£46.00
250mg
Please to purchase this item. Need a web account? Register with us today!
10224842
View Documents
Thermo Scientific Maybridge
CC17412DA
1 g Amber Glass Bottle
£116.00
1g
Please to purchase this item. Need a web account? Register with us today!
10396584
View Documents
Thermo Scientific Maybridge
CC17412DE
5 g Amber Glass Bottle
£530.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

186498-02-2
207.04
WHDIUBHAKZDSJL-UHFFFAOYSA-N
2795359
OB(O)C1=CC=C(C=C1)N1CCOCC1
C10H14BNO3
MFCD03095169
4-morpholinophenylboronic acid, 4-morpholinophenyl boronic acid, 4-morpholino phenylboronic acid, 4-morpholin-4-yl-phenylboronic acid, 4-morpholin-4-ylphenyl boronic acid, 4-4-boronophenyl morpholine, 4-morpholino phenyl boronic acid, 4-morpholin-4-yl phenylboronic acid, boronic acid, 4-4-morpholinyl phenyl, boronic acid, b-4-4-morpholinyl phenyl
(4-morpholin-4-ylphenyl)boronic acid

Specifications

186498-02-2
C10H14BNO3
4-morpholinophenylboronic acid, 4-morpholinophenyl boronic acid, 4-morpholino phenylboronic acid, 4-morpholin-4-yl-phenylboronic acid, 4-morpholin-4-ylphenyl boronic acid, 4-4-boronophenyl morpholine, 4-morpholino phenyl boronic acid, 4-morpholin-4-yl phenylboronic acid, boronic acid, 4-4-morpholinyl phenyl, boronic acid, b-4-4-morpholinyl phenyl
OB(O)C1=CC=C(C=C1)N1CCOCC1
207.04
207.04
4-Morpholinophenylboronic acid
97%
MFCD03095169
WHDIUBHAKZDSJL-UHFFFAOYSA-N
(4-morpholin-4-ylphenyl)boronic acid
2795359
97%
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.