Learn More
4-n-Propoxyaniline, 97%, Thermo Scientific Chemicals
£37.80 - £144.00
Chemical Identifiers
CAS | 4469-80-1 |
---|---|
Molecular Formula | C9H13NO |
Molecular Weight (g/mol) | 151.21 |
MDL Number | MFCD00043621 |
InChI Key | DWOIGSLSPPLRKO-UHFFFAOYSA-N |
Synonym | p-propoxyaniline, benzenamine, 4-propoxy, 4-n-propoxyaniline, 4-propoxyphenyl amine hydrochloride, 4-propoxyphenylamine, hydrochloride, 4-propyloxyaniline, 4-propoxybenzenamine, 4-propoxy-phenylamine |
PubChem CID | 78221 |
IUPAC Name | 4-propoxyaniline |
SMILES | CCCOC1=CC=C(N)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15491238
|
Thermo Scientific Alfa Aesar
H55096.03 |
1 g |
£37.80
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15401248
|
Thermo Scientific Alfa Aesar
H55096.06 |
5 g |
£144.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-n-Propoxyaniline is used as a reactant for preparation of aggrecanase inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-n-Propoxyaniline is used as a reactant for preparation of aggrecanase inhibitors.
Solubility
Slightly soluble (1.3 g/L at 25°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
4469-80-1 | |
151.21 | |
DWOIGSLSPPLRKO-UHFFFAOYSA-N | |
78221 | |
CCCOC1=CC=C(N)C=C1 |
C9H13NO | |
MFCD00043621 | |
p-propoxyaniline, benzenamine, 4-propoxy, 4-n-propoxyaniline, 4-propoxyphenyl amine hydrochloride, 4-propoxyphenylamine, hydrochloride, 4-propyloxyaniline, 4-propoxybenzenamine, 4-propoxy-phenylamine | |
4-propoxyaniline |
Specifications
4469-80-1 | |
110°C (230°F) | |
MFCD00043621 | |
Slightly soluble (1.3g/L at 25°C). | |
CCCOC1=CC=C(N)C=C1 | |
151.21 | |
151.21 | |
Liquid |
Brown | |
C9H13NO | |
p-propoxyaniline, benzenamine, 4-propoxy, 4-n-propoxyaniline, 4-propoxyphenyl amine hydrochloride, 4-propoxyphenylamine, hydrochloride, 4-propyloxyaniline, 4-propoxybenzenamine, 4-propoxy-phenylamine | |
DWOIGSLSPPLRKO-UHFFFAOYSA-N | |
4-propoxyaniline | |
78221 | |
97% | |
4-n-Propoxyaniline |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 224-740-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only