Learn More
Lenz Laborglasintrumente 4 Necks Round Bottom Flasks Side Necks Parallel
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 03344485
 View more versions of this product
Product Code. 11836074
  • £392.00 / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Four-neck round-bottom flasks
  • Side necks parallel
  • Reaction flasks acc. to DIN 12392
  • 3 parallel side necks
  • Conical ground joints
  • Inscription label
  • Material: DURAN™
Specifications
Specifications
236 mm
6 L
Side Neck 29/32 Center Neck 45/40