Learn More
4'-Nitroacetophenone, 98%, Thermo Scientific Chemicals
£22.61 - £217.00
Chemical Identifiers
CAS | 100-19-6 |
---|---|
Molecular Formula | C8H7NO3 |
Molecular Weight (g/mol) | 165.15 |
MDL Number | MFCD00007355 |
InChI Key | YQYGPGKTNQNXMH-UHFFFAOYSA-N |
Synonym | p-nitroacetophenone, 4'-nitroacetophenone, 4-nitroacetophenone, 1-4-nitrophenyl ethanone, p-acetylnitrobenzene, paranitroacetophenone, p-nitrophenyl methyl ketone, 1-4-nitrophenyl ethan-1-one, ethanone, 1-4-nitrophenyl, pnap |
PubChem CID | 7487 |
ChEBI | CHEBI:28735 |
IUPAC Name | 1-(4-nitrophenyl)ethanone |
SMILES | CC(=O)C1=CC=C(C=C1)[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11421568
|
Thermo Scientific Alfa Aesar
A13877.18 |
50 g |
£22.61
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11431568
|
Thermo Scientific Alfa Aesar
A13877.30 |
250 g |
£73.57
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11411568
|
Thermo Scientific Alfa Aesar
A13877.0B |
1000 g | N/A | ||||||
Description
4'-Nitroacetophenone is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4′-Nitroacetophenone is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
Solubility
Insoluble in water.
Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, strong alkalis.
Chemical Identifiers
100-19-6 | |
165.15 | |
YQYGPGKTNQNXMH-UHFFFAOYSA-N | |
7487 | |
1-(4-nitrophenyl)ethanone |
C8H7NO3 | |
MFCD00007355 | |
p-nitroacetophenone, 4'-nitroacetophenone, 4-nitroacetophenone, 1-4-nitrophenyl ethanone, p-acetylnitrobenzene, paranitroacetophenone, p-nitrophenyl methyl ketone, 1-4-nitrophenyl ethan-1-one, ethanone, 1-4-nitrophenyl, pnap | |
CHEBI:28735 | |
CC(=O)C1=CC=C(C=C1)[N+]([O-])=O |
Specifications
100-19-6 | |
201°C to 202°C | |
98% | |
MFCD00007355 | |
p-nitroacetophenone, 4'-nitroacetophenone, 4-nitroacetophenone, 1-4-nitrophenyl ethanone, p-acetylnitrobenzene, paranitroacetophenone, p-nitrophenyl methyl ketone, 1-4-nitrophenyl ethan-1-one, ethanone, 1-4-nitrophenyl, pnap | |
YQYGPGKTNQNXMH-UHFFFAOYSA-N | |
1-(4-nitrophenyl)ethanone | |
7487 | |
165.15 | |
4'-Nitroacetophenone |
77°C to 81°C | |
149°C (300°F) | |
C8H7NO3 | |
607777 | |
Insoluble in water. | |
CC(=O)C1=CC=C(C=C1)[N+]([O-])=O | |
165.15 | |
CHEBI:28735 | |
98% |
Safety and Handling
GHS H Statement
H301
P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
H302-H319-H500
missing translation for 'einecsNumber' : 202-827-4
missing translation for 'rtecsNumber' : AM9627000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- 4'-Nitroacetophenone
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : 4-Nitroacetophenone
RUO – Research Use Only