missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Pyrrolidine-1-sulfonyl)-benzenesulfonyl chloride, Technical Grade, Thermo Scientific™
£42.60 - £261.00
Chemical Identifiers
CAS | 165669-32-9 |
---|---|
Molecular Formula | C10H12ClNO4S2 |
Molecular Weight (g/mol) | 309.779 |
InChI Key | ZXYZKIFFWKXROK-UHFFFAOYSA-N |
Synonym | 4-pyrrolidine-1-sulfonyl-benzenesulfonyl chloride, 4-pyrrolidine-1-sulfonyl benzene-1-sulfonyl chloride, 4-pyrrolidin-1-ylsulfonyl benzenesulfonyl chloride, 4-pyrrolidine-1-sulfonyl benzenesulfonyl chloride, benzenesulfonylchloride, 4-1-pyrrolidinylsulfonyl, 4-pyrrolidin-1-ylsulphonyl benzenesulphonyl chloride, 4-pyrrolidine-1-sulfonyl phenylsulfonyl chloride, benzenesulfonyl chloride, 4-1-pyrrolidinylsulfonyl, 4-pyrrolidin-1-ylsulfonyl benzene-1-sulfonyl chloride |
PubChem CID | 2779793 |
IUPAC Name | 4-pyrrolidin-1-ylsulfonylbenzenesulfonyl chloride |
SMILES | C1CCN(C1)S(=O)(=O)C2=CC=C(C=C2)S(=O)(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10653793
|
Thermo Scientific Maybridge
MO00517CB |
250 mg |
£42.60
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10733253
|
Thermo Scientific Maybridge
MO00517DA |
1 g |
£61.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10410584
|
Thermo Scientific Maybridge
MO00517EA |
10 g |
£261.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
165669-32-9 | |
309.779 | |
4-pyrrolidine-1-sulfonyl-benzenesulfonyl chloride, 4-pyrrolidine-1-sulfonyl benzene-1-sulfonyl chloride, 4-pyrrolidin-1-ylsulfonyl benzenesulfonyl chloride, 4-pyrrolidine-1-sulfonyl benzenesulfonyl chloride, benzenesulfonylchloride, 4-1-pyrrolidinylsulfonyl, 4-pyrrolidin-1-ylsulphonyl benzenesulphonyl chloride, 4-pyrrolidine-1-sulfonyl phenylsulfonyl chloride, benzenesulfonyl chloride, 4-1-pyrrolidinylsulfonyl, 4-pyrrolidin-1-ylsulfonyl benzene-1-sulfonyl chloride | |
4-pyrrolidin-1-ylsulfonylbenzenesulfonyl chloride |
C10H12ClNO4S2 | |
ZXYZKIFFWKXROK-UHFFFAOYSA-N | |
2779793 | |
C1CCN(C1)S(=O)(=O)C2=CC=C(C=C2)S(=O)(=O)Cl |
Specifications
165669-32-9 | |
4-pyrrolidine-1-sulfonyl-benzenesulfonyl chloride, 4-pyrrolidine-1-sulfonyl benzene-1-sulfonyl chloride, 4-pyrrolidin-1-ylsulfonyl benzenesulfonyl chloride, 4-pyrrolidine-1-sulfonyl benzenesulfonyl chloride, benzenesulfonylchloride, 4-1-pyrrolidinylsulfonyl, 4-pyrrolidin-1-ylsulphonyl benzenesulphonyl chloride, 4-pyrrolidine-1-sulfonyl phenylsulfonyl chloride, benzenesulfonyl chloride, 4-1-pyrrolidinylsulfonyl, 4-pyrrolidin-1-ylsulfonyl benzene-1-sulfonyl chloride | |
C1CCN(C1)S(=O)(=O)C2=CC=C(C=C2)S(=O)(=O)Cl | |
309.779 | |
309.79 | |
Technical |
C10H12ClNO4S2 | |
ZXYZKIFFWKXROK-UHFFFAOYSA-N | |
4-pyrrolidin-1-ylsulfonylbenzenesulfonyl chloride | |
2779793 | |
Tech. | |
4-(Pyrrolidine-1-sulfonyl)-benzenesulfonyl chloride |