missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 4-Thiouracil, 97%
£20.98 - £71.00
Chemical Identifiers
CAS | 591-28-6 |
---|---|
Molecular Formula | C4H4N2OS |
Molecular Weight (g/mol) | 128.15 |
MDL Number | MFCD00090842 |
InChI Key | OVONXEQGWXGFJD-UHFFFAOYSA-N |
Synonym | 4-thiouracil, 2-hydroxy-4 1h-pyrimidinethione, 4-thioxo-3,4-dihydropyrimidin-2 1h-one, 2-hydroxy-4-mercaptopyrimidine, uracil, 4-thio, 4-mercaptopyrimidin-2-ol, 4-sulfanylpyrimidin-2-ol, 2-hydroxy-4-thiopyrimidine, 2 1h-pyrimidinone, 3,4-dihydro-4-thioxo, 2 1h-pyrimidinone, 4-mercapto-9ci |
PubChem CID | 2734394 |
IUPAC Name | 4-sulfanylidene-1H-pyrimidin-2-one |
SMILES | O=C1NC=CC(=S)N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10560754
|
Thermo Scientific Chemicals
359932500 |
250 mg | Glass bottle | N/A | |||||
10058981
|
Thermo Scientific Chemicals
359930010 |
1 g | Glass bottle |
£71.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
591-28-6 | |
128.15 | |
OVONXEQGWXGFJD-UHFFFAOYSA-N | |
2734394 | |
O=C1NC=CC(=S)N1 |
C4H4N2OS | |
MFCD00090842 | |
4-thiouracil, 2-hydroxy-4 1h-pyrimidinethione, 4-thioxo-3,4-dihydropyrimidin-2 1h-one, 2-hydroxy-4-mercaptopyrimidine, uracil, 4-thio, 4-mercaptopyrimidin-2-ol, 4-sulfanylpyrimidin-2-ol, 2-hydroxy-4-thiopyrimidine, 2 1h-pyrimidinone, 3,4-dihydro-4-thioxo, 2 1h-pyrimidinone, 4-mercapto-9ci | |
4-sulfanylidene-1H-pyrimidin-2-one |
Specifications
591-28-6 | |
100.0 | |
Yellow | |
96% min. (HPLC) | |
MFCD00090842 | |
OVONXEQGWXGFJD-UHFFFAOYSA-N | |
4-sulfanylidene-1H-pyrimidin-2-one | |
2734394 | |
97% | |
4-Thiouracil, 97% |
96.0 | |
295°C | |
Authentic | |
C4H4N2OS | |
4-thiouracil, 2-hydroxy-4 1h-pyrimidinethione, 4-thioxo-3,4-dihydropyrimidin-2 1h-one, 2-hydroxy-4-mercaptopyrimidine, uracil, 4-thio, 4-mercaptopyrimidin-2-ol, 4-sulfanylpyrimidin-2-ol, 2-hydroxy-4-thiopyrimidine, 2 1h-pyrimidinone, 3,4-dihydro-4-thioxo, 2 1h-pyrimidinone, 4-mercapto-9ci | |
O=C1NC=CC(=S)N1 | |
128.15 | |
128.15 | |
Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Do not eat,drink or smoke when using this product.
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 4-Thiouracil
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
RUO – Research Use Only