missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific Chemicals

Chemical Identifiers

CAS 329-15-7
Molecular Formula C8H4ClF3O
Molecular Weight (g/mol) 208.56
MDL Number MFCD00000694
InChI Key OXZYBOLWRXENKT-UHFFFAOYSA-N
Synonym 4-trifluoromethyl benzoyl chloride, p-trifluoromethyl benzoyl chloride, p-trifluoromethylbenzoyl chloride, benzoyl chloride, 4-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluoyl chloride, 4-trifluoromethyl benzoylchloride, p-trifluoromethyl benzoic acid chloride, ptf-boc, 4-trifluoromethylbenzoylchloride, 4-trifluoromethyl-benzoyl chloride
PubChem CID 67607
IUPAC Name 4-(trifluoromethyl)benzoyl chloride
SMILES FC(F)(F)C1=CC=C(C=C1)C(Cl)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11432237
View Documents
Thermo Scientific Alfa Aesar
A14176.06
5 g N/A
N/A
11442237
View Documents
Thermo Scientific Alfa Aesar
A14176.14
25 g N/A
N/A
11452237
View Documents
Thermo Scientific Alfa Aesar
A14176.22
100 g N/A
N/A
Description

Description

4-(Trifluoromethyl)benzoyl chloride is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzoyl chloride is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store under dry inert gas. Protect from humidity and water. Incompatible with water, bases, alcohol and oxidizing agents.
Specifications

Chemical Identifiers

329-15-7
208.56
OXZYBOLWRXENKT-UHFFFAOYSA-N
67607
FC(F)(F)C1=CC=C(C=C1)C(Cl)=O
C8H4ClF3O
MFCD00000694
4-trifluoromethyl benzoyl chloride, p-trifluoromethyl benzoyl chloride, p-trifluoromethylbenzoyl chloride, benzoyl chloride, 4-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluoyl chloride, 4-trifluoromethyl benzoylchloride, p-trifluoromethyl benzoic acid chloride, ptf-boc, 4-trifluoromethylbenzoylchloride, 4-trifluoromethyl-benzoyl chloride
4-(trifluoromethyl)benzoyl chloride

Specifications

329-15-7
1.404
78°C (172°F)
C8H4ClF3O
MFCD00000694
391282
4-trifluoromethyl benzoyl chloride, p-trifluoromethyl benzoyl chloride, p-trifluoromethylbenzoyl chloride, benzoyl chloride, 4-trifluoromethyl, alpha,alpha,alpha-trifluoro-p-toluoyl chloride, 4-trifluoromethyl benzoylchloride, p-trifluoromethyl benzoic acid chloride, ptf-boc, 4-trifluoromethylbenzoylchloride, 4-trifluoromethyl-benzoyl chloride
OXZYBOLWRXENKT-UHFFFAOYSA-N
4-(trifluoromethyl)benzoyl chloride
67607
98%
-3°C
188°C to 190°C
Pungent
1.477
UN3265
Moisture sensitive
Reacts with water.
FC(F)(F)C1=CC=C(C=C1)C(Cl)=O
208.56
208.57
4-(Trifluoromethyl)benzoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 206-342-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only