missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trimethylsilyl)diphenylacetylene, 99%, Thermo Scientific Chemicals
£21.40 - £219.00
Chemical Identifiers
CAS | 136459-72-8 |
---|---|
Molecular Formula | C17H18Si |
Molecular Weight (g/mol) | 250.416 |
MDL Number | MFCD09953470 |
InChI Key | UZRJWRFLYPENFH-UHFFFAOYSA-N |
Synonym | 4-trimethylsilyl diphenylacetylene, trimethyl 4-phenylethynyl phenyl silane, trimethyl 4-2-phenylethynyl phenyl silane, acmc-20ajdj, 4-phenylethynyl phenyltrimethylsilane, trimethyl-4-2-phenylethynyl phenyl silane, 1-2-phenylethynyl-4-trimethylsilyl benzene, 1-phenyl-2-p-trimethylsilyl phenyl acetylene |
PubChem CID | 21093331 |
IUPAC Name | trimethyl-[4-(2-phenylethynyl)phenyl]silane |
SMILES | C[Si](C)(C)C1=CC=C(C=C1)C#CC2=CC=CC=C2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11461150
|
Thermo Scientific Alfa Aesar
H26300.03 |
1 g |
£21.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11471150
|
Thermo Scientific Alfa Aesar
H26300.06 |
5 g |
£63.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11481150
|
Thermo Scientific Alfa Aesar
H26300.14 |
25 g |
£219.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
136459-72-8 | |
250.416 | |
UZRJWRFLYPENFH-UHFFFAOYSA-N | |
21093331 | |
C[Si](C)(C)C1=CC=C(C=C1)C#CC2=CC=CC=C2 |
C17H18Si | |
MFCD09953470 | |
4-trimethylsilyl diphenylacetylene, trimethyl 4-phenylethynyl phenyl silane, trimethyl 4-2-phenylethynyl phenyl silane, acmc-20ajdj, 4-phenylethynyl phenyltrimethylsilane, trimethyl-4-2-phenylethynyl phenyl silane, 1-2-phenylethynyl-4-trimethylsilyl benzene, 1-phenyl-2-p-trimethylsilyl phenyl acetylene | |
trimethyl-[4-(2-phenylethynyl)phenyl]silane |
Specifications
37°C to 39°C | |
C17H18Si | |
4-trimethylsilyl diphenylacetylene, trimethyl 4-phenylethynyl phenyl silane, trimethyl 4-2-phenylethynyl phenyl silane, acmc-20ajdj, 4-phenylethynyl phenyltrimethylsilane, trimethyl-4-2-phenylethynyl phenyl silane, 1-2-phenylethynyl-4-trimethylsilyl benzene, 1-phenyl-2-p-trimethylsilyl phenyl acetylene | |
C[Si](C)(C)C1=CC=C(C=C1)C#CC2=CC=CC=C2 | |
250.416 | |
250.42 | |
4-(Trimethylsilyl)diphenylacetylene |
136459-72-8 | |
MFCD09953470 | |
UZRJWRFLYPENFH-UHFFFAOYSA-N | |
trimethyl-[4-(2-phenylethynyl)phenyl]silane | |
21093331 | |
99% |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only