Learn More
RETSCH 400 dia. x 65 mmH Stainless Steel Test Sieve, ISO Certified
£273.00 - £990.00
Specifications
Diameter (Metric) Outer | 400 mm |
---|---|
Frame Material | Stainless Steel |
Mesh Material | Stainless Steel |
For Use With (Application) | For separation, fractioning, particle size determination |
Quantity | 1Each |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
10781623
|
RETSCH
60.166.100000 |
100 mm |
£441.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10514882
|
RETSCH
60.166.106000 |
106 mm |
£441.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10145981
|
RETSCH
60.166.000100 |
100 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10370862
|
RETSCH
60.166.112000 |
112 mm |
£441.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10488921
|
RETSCH
60.166.000106 |
106 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10546222
|
RETSCH
60.166.000112 |
112 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10750083
|
RETSCH
60.166.125000 |
125 mm |
£441.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10620492
|
RETSCH
60.166.000125 |
125 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10187021
|
RETSCH
60.166.000140 |
140 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10630462
|
RETSCH
60.166.000150 |
150 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10600292
|
RETSCH
60.166.000160 |
160 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10248691
|
RETSCH
60.166.000180 |
180 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10166791
|
RETSCH
60.166.000200 |
200 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10486811
|
RETSCH
60.166.000212 |
212 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10268831
|
RETSCH
60.166.000224 |
224 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10064921
|
RETSCH
60.166.000250 |
250 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10791223
|
RETSCH
60.166.000025 |
25 μm |
£990.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10670462
|
RETSCH
60.166.000280 |
280 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10447201
|
RETSCH
60.166.000315 |
315 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10419661
|
RETSCH
60.166.000032 |
32 μm |
£665.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10390812
|
RETSCH
60.166.000300 |
300 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10300822
|
RETSCH
60.166.000355 |
355 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10525252
|
RETSCH
60.166.000036 |
36 μm |
£600.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10258581
|
RETSCH
60.166.000038 |
38 μm |
£580.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10360862
|
RETSCH
60.166.000400 |
400 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10721613
|
RETSCH
60.166.000040 |
40 μm |
£427.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10749892
|
RETSCH
60.166.000425 |
425 μm |
£273.00
Each |
Estimated Shipment: 14-11-2024
Log in to see stock. |
|||||
10258731
|
RETSCH
60.166.000450 |
450 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10136591
|
RETSCH
60.166.000045 |
45 μm |
£412.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10390242
|
RETSCH
60.166.000500 |
500 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10369601
|
RETSCH
60.166.000050 |
50 μm |
£341.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10218681
|
RETSCH
60.166.000053 |
53 μm |
£320.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10593332
|
RETSCH
60.166.000560 |
560 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10761813
|
RETSCH
60.166.000056 |
56 μm |
£304.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10740083
|
RETSCH
60.166.000600 |
600 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10563912
|
RETSCH
60.166.000630 |
630 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10438351
|
RETSCH
60.166.000063 |
63 μm |
£296.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10268921
|
RETSCH
60.166.000710 |
710 μm |
£273.00
Each |
Estimated Shipment: 05-12-2024
Log in to see stock. |
|||||
10711053
|
RETSCH
60.166.071000 |
71 mm |
£441.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10749322
|
RETSCH
60.166.000071 |
71 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10167071
|
RETSCH
60.166.075000 |
75 mm |
£441.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10516032
|
RETSCH
60.166.000075 |
75 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10700473
|
RETSCH
60.166.000800 |
800 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10034981
|
RETSCH
60.166.000080 |
80 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10105981
|
RETSCH
60.166.000850 |
850 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10340722
|
RETSCH
60.166.080000 |
80 mm |
£441.00
Each |
Estimated Shipment: 14-11-2024
Log in to see stock. |
|||||
10668571
|
RETSCH
60.166.000900 |
900 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10147171
|
RETSCH
60.166.090000 |
90 mm |
£441.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
10670282
|
RETSCH
60.166.000090 |
90 μm |
£273.00
Each |
Estimated Shipment: 19-12-2024
Log in to see stock. |
|||||
Specifications
400 mm | |
Stainless Steel | |
1Each | |
ISO 3310/1 | |
Shaker AS 450 |
Stainless Steel | |
For separation, fractioning, particle size determination | |
Test Sieve | |
Stainless Steel | |
65mm |