missing translation for 'onlineSavingsMsg'
Learn More

5-(3-Trifluoromethoxyphenyl)-1H-tetrazole, 97%, Thermo Scientific™

£42.30 - £156.00

Chemical Identifiers

CAS 216144-09-1
Molecular Formula C8H5F3N4O
Molecular Weight (g/mol) 230.15
MDL Number MFCD01075683
InChI Key YKYWVXHVSVSQOX-UHFFFAOYSA-N
Synonym 5-3-trifluoromethoxy phenyl-1h-tetrazole, 5-3-trifluoromethoxy phenyl-2h-tetrazole, 5-3-trifluoromethoxyphenyl-1h-tetrazole, 5-3-trifluoromethoxy phenyl-1h-1,2,3,4-tetrazole, 3-1h-1,2,3,4-tetraazol-5-yl phenoxy trifluoromethane, acmc-1cs77, 5-3-trifluoromethoxy phenyl tetrazole, 2h-tetrazole,5-3-trifluoromethoxy phenyl, 5-3-trifluoromethyloxy phenyl-2h-1,2,3,4-tetrazole
PubChem CID 2777350
IUPAC Name 5-[3-(trifluoromethoxy)phenyl]-2H-tetrazole
SMILES C1=CC(=CC(=C1)OC(F)(F)F)C2=NNN=N2
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11318236
View Documents
Thermo Scientific Alfa Aesar
L15268.MC
100 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11308236
View Documents
Thermo Scientific Alfa Aesar
L15268.ME
500 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

216144-09-1
230.15
YKYWVXHVSVSQOX-UHFFFAOYSA-N
2777350
C1=CC(=CC(=C1)OC(F)(F)F)C2=NNN=N2
C8H5F3N4O
MFCD01075683
5-3-trifluoromethoxy phenyl-1h-tetrazole, 5-3-trifluoromethoxy phenyl-2h-tetrazole, 5-3-trifluoromethoxyphenyl-1h-tetrazole, 5-3-trifluoromethoxy phenyl-1h-1,2,3,4-tetrazole, 3-1h-1,2,3,4-tetraazol-5-yl phenoxy trifluoromethane, acmc-1cs77, 5-3-trifluoromethoxy phenyl tetrazole, 2h-tetrazole,5-3-trifluoromethoxy phenyl, 5-3-trifluoromethyloxy phenyl-2h-1,2,3,4-tetrazole
5-[3-(trifluoromethoxy)phenyl]-2H-tetrazole

Specifications

216144-09-1
C8H5F3N4O
5-3-trifluoromethoxy phenyl-1h-tetrazole, 5-3-trifluoromethoxy phenyl-2h-tetrazole, 5-3-trifluoromethoxyphenyl-1h-tetrazole, 5-3-trifluoromethoxy phenyl-1h-1,2,3,4-tetrazole, 3-1h-1,2,3,4-tetraazol-5-yl phenoxy trifluoromethane, acmc-1cs77, 5-3-trifluoromethoxy phenyl tetrazole, 2h-tetrazole,5-3-trifluoromethoxy phenyl, 5-3-trifluoromethyloxy phenyl-2h-1,2,3,4-tetrazole
C1=CC(=CC(=C1)OC(F)(F)F)C2=NNN=N2
230.15
230.15
5-(3-Trifluoromethoxyphenyl)-1H-tetrazole
156°C to 159°C
MFCD01075683
YKYWVXHVSVSQOX-UHFFFAOYSA-N
5-[3-(trifluoromethoxy)phenyl]-2H-tetrazole
2777350
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications