Learn More
5-Acetyl-2-methoxypyridine, 97%, Thermo Scientific Chemicals
£54.40 - £173.00
Chemical Identifiers
CAS | 213193-32-9 |
---|---|
Molecular Formula | C8H9NO2 |
Molecular Weight (g/mol) | 151.17 |
InChI Key | RYOQZXOVBJIUSX-UHFFFAOYSA-N |
Synonym | 5-acetyl-2-methoxypyridine, 1-6-methoxypyridin-3-yl ethanone, 1-6-methoxypyridin-3-yl ethan-1-one, 1-6-methoxy-3-pyridyl ethanone, ethanone, 1-6-methoxy-3-pyridinyl, 3-acetyl-6-methoxypyridine, 1-6-methoxy-3-pyridinyl ethanone, pubchem16770, acmc-209fj4, 2-methoxy-5-acetylpyridine |
PubChem CID | 10534837 |
IUPAC Name | 1-(6-methoxypyridin-3-yl)ethanone |
SMILES | CC(=O)C1=CN=C(C=C1)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11986381
|
Thermo Scientific Acros
438170010 |
1 g |
£54.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11916391
|
Thermo Scientific Acros
438170050 |
5 g |
£173.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
213193-32-9 | |
151.17 | |
5-acetyl-2-methoxypyridine, 1-6-methoxypyridin-3-yl ethanone, 1-6-methoxypyridin-3-yl ethan-1-one, 1-6-methoxy-3-pyridyl ethanone, ethanone, 1-6-methoxy-3-pyridinyl, 3-acetyl-6-methoxypyridine, 1-6-methoxy-3-pyridinyl ethanone, pubchem16770, acmc-209fj4, 2-methoxy-5-acetylpyridine | |
1-(6-methoxypyridin-3-yl)ethanone |
C8H9NO2 | |
RYOQZXOVBJIUSX-UHFFFAOYSA-N | |
10534837 | |
CC(=O)C1=CN=C(C=C1)OC |
Specifications
213193-32-9 | |
100.0 | |
White to Yellow | |
97% | |
C8H9NO2 | |
RYOQZXOVBJIUSX-UHFFFAOYSA-N | |
1-(6-methoxypyridin-3-yl)ethanone | |
10534837 | |
97% | |
5-Acetyl-2-methoxypyridine |
96.0 | |
53.0°C to 54.0°C | |
Authentic | |
Glass bottle | |
5-acetyl-2-methoxypyridine, 1-6-methoxypyridin-3-yl ethanone, 1-6-methoxypyridin-3-yl ethan-1-one, 1-6-methoxy-3-pyridyl ethanone, ethanone, 1-6-methoxy-3-pyridinyl, 3-acetyl-6-methoxypyridine, 1-6-methoxy-3-pyridinyl ethanone, pubchem16770, acmc-209fj4, 2-methoxy-5-acetylpyridine | |
CC(=O)C1=CN=C(C=C1)OC | |
151.17 | |
151.17 | |
Crystalline Powder or Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
- 5-Acetyl-2-methoxypyridine
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only