Learn More
5-Chloro-2-mercaptobenzothiazole, 98%, Thermo Scientific Chemicals
£44.42 - £126.63
Chemical Identifiers
CAS | 5331-91-9 |
---|---|
Molecular Formula | C7H4ClNS2 |
Molecular Weight (g/mol) | 201.69 |
MDL Number | MFCD00005783 |
InChI Key | NKYDKCVZNMNZCM-UHFFFAOYSA-N |
Synonym | 5-chloro-2-mercaptobenzothiazole, 5-chloro-1,3-benzothiazole-2-thiol, 5-chlorobenzo d thiazole-2 3h-thione, sh-benzothiazole, 5-chloro-2-benzothiazolethiol, 2 3h-benzothiazolethione, 5-chloro, 5-chlorobenzo d thiazole-2-thiol, 5-chlorobenzothiazole-2-thiol, benzothiazole, 5-chloro-2-mercapto, 2-benzothiazolethiol, 5-chloro |
PubChem CID | 2723842 |
IUPAC Name | 5-chloro-3H-1,3-benzothiazole-2-thione |
SMILES | ClC1=CC=C2SC(=S)NC2=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11492758
|
Thermo Scientific Alfa Aesar
A16747.06 |
5 g |
£44.42
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11402768
|
Thermo Scientific Alfa Aesar
A16747.14 |
25 g |
£126.63
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5331-91-9 | |
201.69 | |
NKYDKCVZNMNZCM-UHFFFAOYSA-N | |
2723842 | |
ClC1=CC=C2SC(=S)NC2=C1 |
C7H4ClNS2 | |
MFCD00005783 | |
5-chloro-2-mercaptobenzothiazole, 5-chloro-1,3-benzothiazole-2-thiol, 5-chlorobenzo d thiazole-2 3h-thione, sh-benzothiazole, 5-chloro-2-benzothiazolethiol, 2 3h-benzothiazolethione, 5-chloro, 5-chlorobenzo d thiazole-2-thiol, 5-chlorobenzothiazole-2-thiol, benzothiazole, 5-chloro-2-mercapto, 2-benzothiazolethiol, 5-chloro | |
5-chloro-3H-1,3-benzothiazole-2-thione |
Specifications
5331-91-9 | |
C7H4ClNS2 | |
5-chloro-2-mercaptobenzothiazole, 5-chloro-1,3-benzothiazole-2-thiol, 5-chlorobenzo d thiazole-2 3h-thione, sh-benzothiazole, 5-chloro-2-benzothiazolethiol, 2 3h-benzothiazolethione, 5-chloro, 5-chlorobenzo d thiazole-2-thiol, 5-chlorobenzothiazole-2-thiol, benzothiazole, 5-chloro-2-mercapto, 2-benzothiazolethiol, 5-chloro | |
ClC1=CC=C2SC(=S)NC2=C1 | |
201.69 | |
201.7 | |
5-Chloro-2-mercaptobenzothiazole |
198°C to 200°C | |
MFCD00005783 | |
NKYDKCVZNMNZCM-UHFFFAOYSA-N | |
5-chloro-3H-1,3-benzothiazole-2-thione | |
2723842 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'einecsNumber' : 226-235-0
missing translation for 'rtecsNumber' : DL6490000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- 5-Chloro-2-mercaptobenzothiazole
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 5-Chloro-2-mercaptobenzothiazole
RUO – Research Use Only