missing translation for 'onlineSavingsMsg'
Learn More

5-Methylcyclohexane-1,3-dione, 98%, Thermo Scientific Chemicals

£27.94 - £283.41

Chemical Identifiers

CAS 4341-24-6
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00010379
InChI Key DMIIMPQQPXUKOO-UHFFFAOYSA-N
PubChem CID 458095
IUPAC Name 5-methylcyclohexane-1,3-dione
SMILES CC1CC(=O)CC(=O)C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11422348
View Documents
Thermo Scientific Alfa Aesar
A15280.03
1 g
£27.94
1g
Please to purchase this item. Need a web account? Register with us today!
11432348
View Documents
Thermo Scientific Alfa Aesar
A15280.06
5 g
£82.41
5g
Please to purchase this item. Need a web account? Register with us today!
11442348
View Documents
Thermo Scientific Alfa Aesar
A15280.14
25 g
£283.41
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4341-24-6
126.155
DMIIMPQQPXUKOO-UHFFFAOYSA-N
5-methylcyclohexane-1,3-dione
C7H10O2
MFCD00010379
458095
CC1CC(=O)CC(=O)C1

Specifications

4341-24-6
C7H10O2
1099692
DMIIMPQQPXUKOO-UHFFFAOYSA-N
5-methylcyclohexane-1,3-dione
458095
98%
126°C to 130°C
MFCD00010379
5-methyl-1,3-cyclohexanedione, 1,3-cyclohexanedione, 5-methyl, 5-methyl-cyclohexane-1,3-dione, pubchem14279, acmc-1aftb, 5-methylcyclohexane dione, 1,3-dioxo-5-methylcyclohexane, 1,3-cyclohexanedione,5-methyl, 5-methyl cyclohexane-1,3-dione
CC1CC(=O)CC(=O)C1
126.155
126.16
5-Methylcyclohexane-1,3-dione
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only