missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 5-Nitroindoline, 97%
£187.00
Chemical Identifiers
CAS | 32692-19-6 |
---|---|
Molecular Formula | C8H8N2O2 |
Molecular Weight (g/mol) | 164.164 |
MDL Number | MFCD00005709 |
InChI Key | WJQWYAJTPPYORB-UHFFFAOYSA-N |
Synonym | 5-nitroindoline, 1h-indole, 2,3-dihydro-5-nitro, indoline, 5-nitro, 2,3-dihydro-5-nitro-1h-indole, ccris 3306, 5-nitro-2,3-dihydro-indole, 5-nitroindolin, 5-nitro indoline, pubchem7442, acmc-1aera |
PubChem CID | 36219 |
IUPAC Name | 5-nitro-2,3-dihydro-1H-indole |
SMILES | C1CNC2=C1C=C(C=C2)[N+](=O)[O-] |
Description
Chemical Identifiers
32692-19-6 | |
164.164 | |
WJQWYAJTPPYORB-UHFFFAOYSA-N | |
36219 | |
C1CNC2=C1C=C(C=C2)[N+](=O)[O-] |
C8H8N2O2 | |
MFCD00005709 | |
5-nitroindoline, 1h-indole, 2,3-dihydro-5-nitro, indoline, 5-nitro, 2,3-dihydro-5-nitro-1h-indole, ccris 3306, 5-nitro-2,3-dihydro-indole, 5-nitroindolin, 5-nitro indoline, pubchem7442, acmc-1aera | |
5-nitro-2,3-dihydro-1H-indole |
Specifications
32692-19-6 | |
C8H8N2O2 | |
5-nitroindoline, 1h-indole, 2,3-dihydro-5-nitro, indoline, 5-nitro, 2,3-dihydro-5-nitro-1h-indole, ccris 3306, 5-nitro-2,3-dihydro-indole, 5-nitroindolin, 5-nitro indoline, pubchem7442, acmc-1aera | |
C1CNC2=C1C=C(C=C2)[N+](=O)[O-] | |
164.164 | |
164.16 | |
5-Nitroindoline |
92°C to 94°C | |
MFCD00005709 | |
WJQWYAJTPPYORB-UHFFFAOYSA-N | |
5-nitro-2,3-dihydro-1H-indole | |
36219 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 251-158-4
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Alfa Aesar™ 5-Nitroindoline, 97%