missing translation for 'onlineSavingsMsg'
Learn More

5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 132860025

Code : 29

Additional Details : CAS Number : 5965-83-3 Weight : 2.55000kg Transport : UN number : 2585 Chem class : 8 Pack group : III

 View more versions of this product

Product Code. 10403645

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Specifications

5-Sulfosalicylic acid dihydrate
97.5
106.0°C to 110.0°C
Authentic
Plastic bottle
MFCD00007508,MFCD00149540
11, 411
15, 9094
Solubility in water: 127g/L (20°C). Other solubilities: soluble in alcohol and ether
C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
250.27
254.22
Crystalline Powder or Crystals
5965-83-3,97-05-2
100.0
White
98%
C9H14O6S
2.5 kg
01,1118
2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
NFYHZVWMQHQKRU-UHFFFAOYSA-N
2-hydroxy-5-sulfobenzoic acid;dihydrate
2723734
98%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Product Certifications
Promotions

Promotions

RUO – Research Use Only