Learn More
5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals
£29.45 - £129.00
Specifications
Chemical Name or Material | 5-Sulfosalicylic acid dihydrate |
---|---|
CAS | 5965-83-3, 97-05-2 |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 106.0°C to 110.0°C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10275280
|
Thermo Scientific Acros
132862500 |
250 g | Plastic bottle |
£29.45
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10644612
|
Thermo Scientific Acros
132860010 |
1 kg | Plastic bottle |
£76.58
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10403645
|
Thermo Scientific Acros
132860025 |
2.5 kg | Plastic bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
5-Sulfosalicylic acid dihydrate | |
97.5 | |
106.0°C to 110.0°C | |
Authentic | |
Plastic bottle | |
MFCD00007508,MFCD00149540 | |
01,1118 | |
2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd | |
NFYHZVWMQHQKRU-UHFFFAOYSA-N | |
250.27 | |
254.22 | |
Crystalline Powder or Crystals |
5965-83-3, 97-05-2 | |
100.0 | |
White | |
98% | |
C9H14O6S | |
11, 411 | |
15, 9094 | |
Solubility in water: 127g/L (20°C). Other solubilities: soluble in alcohol and ether | |
C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O | |
2723734 | |
98% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
- 5-Sulfosalicylic acid dihydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : 5-Sulfosalicylic acid dihydrate
RUO – Research Use Only