Learn More
Value Plastics 500 Series Hose Barb Fittings, Straight Reducer
A secure tubing connection with semi-rigid tubing
Brand: Value Plastics 50120-93
This item is not returnable.
View return policy
Description
Quick and easy assembly and disassembly without the need for tools. Barbed ridges provide secure grip on the inside of fle in., ible tubing. Constructed of USP compliant ADCF polypropyleneSpecifications
ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP | |
Hose Barb Reducer | |
Straight |
25/Pk. | |
121°C |
Product Content Correction
Product Title
Value Plastics 500 Series Hose Barb Fittings, Straight Reducer > 25 Hose Barb Straight Reducer Fitting PP 1/4 in.1/8
Spot an opportunity for improvement?Share a Content Correction