missing translation for 'onlineSavingsMsg'
Learn More

5000 ppm Chemical Oxygen Demand Standard, Reagecon
BIOPHARMA

Can be used to calibrate, control, verify or validate all COD test methods.

Brand:  Reagecon COD5000

Product Code. 11798515

  • £103.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Chemical Oxygen Demand Reference Standard, Chemical Oxygen Demand Reference Material
  • Can be used to calibrate, control, verify or validate all COD test methods.
  • Yields an accurate test result of 5000 ppm.
  • Useful for achieving COD analytical confidence.
Specifications

Specifications

HDPE Bottle GL45
Liquid
COD Test Methods
500 mL
500 ppm COD
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5000 ppm Chemical Oxygen Demand Standard, Reagecon > 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.