missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ iBind™ Flex Western Starter Kit
Allows better antigen-antibody interaction and solutions flow between the iBind™ Card and membrane for maximum interaction between protein and antibody
Brand: Invitrogen™ SLF2000S
1835.40 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.
This item is not returnable.
View return policy
Description
The iBind™ Flex Western System offers automated western blot processing and can accommodate various blot formats including midi, mini, and vertically cut strips. Based on Sequential Lateral Flow (SLF) technology, all blocking, antibody incubation and washing steps during immunodetection are hands-free. Compared to manual methods, the iBind Flex Western System requires less primary antibody, enables reproducible results and is compatible with downstream detection protocols.The Invitrogen™ iBind™ Flex Western System offers:
- Flexible options—optimize antibody use, change formats and reduce hands-on time
- Cost savings—use up to 80% less primary antibody
- Reproducibility—automated processing provides improved blot-to-blot consistency

Specifications
Starter Kit | |
• 1 iBind Flex Western Device • 10 iBind Flex Cards, store at room temperature. • 1 iBind Flex Solution Kit, store at 2–8°C. |
|
∼2.5 hours | |
Chromogenic, Chemiluminescence, Fluorescence | |
iBind™ |
one midi blot, six vertically-cut strip blots, two mini blots | |
10 iBind cards, 1 iBind solution kit, Western device | |
Room Temperature | |
Nitrocellulose, PVDF | |
1 kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ iBind™ Flex Western Starter Kit > iBind Flex Western Starter Kit
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction