Learn More
Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, EX 2%
Designed for fast and convenient E-Gel™ agarose gel electrophoresis, from DNA sample loading to gel image. Includes E-Gel EX 2% agarose gels.
Brand: Invitrogen™ G8142ST
1189.39 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Includes: E-Gel Power Snap Electrophoresis device,orange viewing glasses,E-Gel Go! agarose gel adaptor,region specific cable with power adaptor.
Description
The Invitrogen E-Gel Power Snap Electrophoresis Device Starter Kit, EX 2%, provides everything you need to quickly start using precast E-Gel EX 2% agarose gels on the device.
The kit includes:
- 10 E-Gel EX agarose gels, 2% (Cat. No. G401002)
- E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
- E-Gel 50 bp DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488099)
- Gel opening knife (Cat. No. EI9010)
Services
1 year warranty

Specifications
• 1 electrophoresis device • 10 E-Gel EX 2% gels • 1 vial E-Gel 50 bp DNA Ladder • 1 power adapter • 1 pair of viewing glasses • 1 E-Gel Go! Adapter • 1 knife Store at room temperature. |
|
1 kit | |
48 VDC |
Electrophoresis System Starter Kit | |
Room Temperature | |
240 VAC |
Product Suggestions
Customers who viewed this item also viewed
Videos
-
Invitrogen E-Gel Power Snap electrophoresis system - How to run E-Gel EX
Special Offers
-
Multiple Manufacturers
More Discovery, Less Cost
Save on thousands of eligible products.Promo Code: 24111Expires: 2025-03-21
Days
3Hours
21Minutes
43Seconds
12New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.