missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Chromogenic Endotoxin Quant Kit

Chromogenic Endotoxin Quant Kits are highly sensitive endpoint assays that measure and detect endotoxin (LPS) in protein, peptide, nucleic acid, or antibody samples using the Amebocyte Lysate Assay from 0.01–1.0 EU/mL. Kits are available in reaction sizes ranging from 30 to 240.

Brand:  Thermo Scientific™ A39552S

220.40 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 15955590

  • £241.00 / 30 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

• Lyophilized amebocyte lysate
• E.coli endotoxin standard
• Chromogenic substrate
• Endotoxin-free water

Store at 2–8°C upon receipt.

0.01 EU/mL
Colorimetric
Pierce™
Endotoxins
30 microplate-well assays containing 0.01 to 1 endotoxin units/mL
0.01 to 0.1 EU/mL, 0.1 to 1.0 EU/mL
End-point Chromogenic Assay
Wet Ice
30 Reactions
Endotoxin Quantitation Kit
Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number0B96160100 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog NumberA39552S
Lot Number0B65F00100 Certificate TypeCertificate of Analysis Date08/08/2024 Catalog NumberA39552S
Lot Number0CC8090100 Certificate TypeCertificate of Analysis Date25/05/2024 Catalog NumberA39552S
Lot Number0CA7X30700 Certificate TypeCertificate of Analysis Date07/04/2024 Catalog NumberA39552S
Lot Number0B13R00700 Certificate TypeCertificate of Analysis Date26/03/2024 Catalog NumberA39552S
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Chromogenic Endotoxin Quant Kit > 30 reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.