Learn More
Thermo Scientific™ Pierce™ High Capacity Endotoxin Removal Spin Columns, 1 mL
Shop All Thermo Scientific ProductsDescription
Reduce endotoxin levels in protein samples by greater than 99% in one hour while maximizing protein recovery with Thermo Scientific™ Pierce High Capacity Endotoxin Removal Resin.
Highlights:
High capacity bind up to 2,000,000 EU/mL to eliminate at least 99% of endotoxins
Durable re-use resin up to 10 times with no loss in performance
Selective recover at least 85% of your protein sample
High performance compatible with FDA guidelines by reducing final EU concentration to less than 5 EU/mL
Fast our new spin cup format enables endotoxin depletion within 1 hour
Cleansingle-use spin columns avoid cross contamination of samples
Optimized spin cups are optimized for different sample volumes
Economical large-volume discounts available
Requires:
Endotoxin-free water
Recommended for:
Eliminate endotoxins from protein or antibody samples prior to cell transfection or animal injection
Specifications
Specifications
| Capacity (Metric) | Up to 2,000,000 EU/mL |
| Formulation | 22mL-capacity centrifuge columns containing 1mL resin |
| Content And Storage | Upon receipt store at 4°C. |
| Form | Liquid |
| Product Line | Pierce |
| Quantity | 5 Columns |
| Target | Endotoxin Removal |
| Type | Spin Column |
| Stationary Phase | Proprietary Ligand |
| Format | Spin Column |
| Show More |
Safety and Handling
- Pierce High Capacity Endotoxin Removal Spin Columns, 1 mL
- Danger
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Reproductive toxicity Category 1
- Skin corrosion/irritation Category 2
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H340-May cause genetic defects.
- H351-Suspected of causing cancer.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.