missing translation for 'onlineSavingsMsg'
Learn More

Jenway™ 6305 UV/Visible Spectrophotometer

Suitable for applications that require measurements in the UV-visible range. Jenway™ 6305 UV/Visible Spectrophotometer includes multi-parameter displays, full interfacing capability, and versatile sampling accessories.

Brand:  Jenway™ 99968-68

3933.00 GBP valid until 2025-12-31
Use promo code "24356" to get your promotional price.


Product Code. 11400539

  • £4140.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Suitable for applications that require measurements in the UV-visible range.
  • Easy operation
  • Multi-parameter displays
  • Full interfacing capability (including to PC)
  • Versatile sampling accessories
  • Three year warranty
  • Supplied with mains lead, pack of 100 disposable cuvettes, 10mm x 10mm cell holder, interface cable, PC application software on CD-ROM (requires free online registration) and operating instructions.
Specifications

Specifications

-300 to 1999
Xenon
UV-Vis Spectrophotometer
198 to 1000 nm
160 mm
<0.001A at 0A at 400nm
<0.5%T at 220 and 340 nm
±2 nm
365 mm
272 mm
8 nm, 6 nm over the UV range
115/230 V
115/230V, 50/60 Hz
50/60 Hz
0.1%T, 0.001 A
<50 W
6 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Jenway™ 6305 UV/Visible Spectrophotometer > Height: 160mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.