missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs Spectra/Por™ 7 10,000 D MWCO Standard RC Pre-Treated Dialysis Kits
With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds
Brand: Spectrum™ 132 117T
Code : NEW
Additional Details : Weight : 0.10000kg
Includes:
- 1meter of pre-treated tubing, a pair of closures and 5 opening picks
Description
- Hydrophilic, symmetric porositiy
- Equilibrated in water
- Packaged wet in preservative solution of 0.05% sodium azide
- For small experiments or qualifying a membrane for a particular application
Specifications
10 kDa | |
7.5 mm | |
0.45 mL/cm vol./length | |
1 m Tubing/Kit |