missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs Spectra/Por™ 7 10,000 D MWCO Standard RC Pre-Treated Dialysis Kits
With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds
£144.49 - £194.00
Specifications
MWCO | 10 kDa |
---|---|
Quantity | 1 m Tubing/Kit |
Includes:
- 1meter of pre-treated tubing, a pair of closures and 5 opening picks
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
15340702
|
Spectrum™
132120T |
6.4 mL/cm vol./length | 29 mm |
£146.00
1m |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15330702
|
Spectrum™
132119T |
3.3 mL/cm vol./length | 20.4 mm |
£194.00
1m |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15320702
|
Spectrum™
132 118T |
1.8 mL/cm vol./length | 15 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15310702
|
Spectrum™
132 117T |
0.45 mL/cm vol./length | 7.5 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Hydrophilic, symmetric porositiy
- Equilibrated in water
- Packaged wet in preservative solution of 0.05% sodium azide
- For small experiments or qualifying a membrane for a particular application
Specifications
10 kDa | |
1 m Tubing/Kit |