missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs Spectra/Por™ 3 3.5 kD MWCO Standard RC Dry Dialysis Kits
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
Brand: Spectrum™ 132725T
137.47 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Includes:
- 1m or 5m of Spectra/Por 3 dialysis tubing
- 1 Standard Spectra/Por Closure (for buoyance)
- 1 weighted Spectra/Por Closure (for orientation)
- 2 adhesive tubing openers
Description
- For basic dialysis applications, or evaluating dialysis tubing for dialysis application
- Packaged dry with glycerin (soak in water) or without glycerin (no soaking required)
Specifications
Trial Kit | |
Regenerated Cellulose | |
0.2 in. | |
5 m | |
9.3 mL | |
5 m Tubing/Kit |
3.5 kDa | |
34 mm | |
16 ft. | |
Autoclavable | |
5m roll of Spectra/Por™ 1, 2 or 3 dialysis tubing, 1 standard spectra/por closure (aids buoyancy), 1 weighted spectra/por closure (aids vertical orientation), 5 opening picks | |
ideal for small volume dialysis or evaluation of membrane for an application |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spectrum™ Labs Spectra/Por™ 3 3.5 kD MWCO Standard RC Dry Dialysis Kits > MWCO: 3.5 kd; For Spectra/Por 3; Flat W: 54mm; Dia.: 34mm; Closure: 75mm; Vol./L: 9.3mL/cm
Spot an opportunity for improvement?Share a Content Correction