missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs Spectra/Por™ 3 3.5 kD MWCO Standard RC Dry Dialysis Kits
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
£44.55 - £223.00
Specifications
MWCO | 3.5 kDa |
---|
Includes:
- 1m or 5m of Spectra/Por 3 dialysis tubing
- 1 Standard Spectra/Por Closure (for buoyance)
- 1 weighted Spectra/Por Closure (for orientation)
- 2 adhesive tubing openers
Product Code | Brand | Length (Metric) | Diameter (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Quantity | Price | Quantity & Availability | |||
15360772
|
Spectrum™
132724T |
6.4 mL/cm vol./length | 29 mm | 5 m Tubing/Kit |
£223.00
5m |
Estimated Shipment: 17-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11571060
|
Spectrum™
132725T |
5 m | 34 mm | 5 m Tubing/Kit |
£223.00
5m |
Estimated Shipment: 17-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15340772
|
Spectrum™
132 720T1 |
1.1 mL/cm vol./length | 11.5 mm | 1 m Tubing/Kit |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15370772
|
Spectrum™
132 724T1 |
6.4 mL/cm vol./length | 29 mm | 1 m Tubing/Kit |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
15380772
|
Spectrum™
132 725T1 |
9.3 mL/cm vol./length | 34 mm | 1 m Tubing/Kit |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- For basic dialysis applications, or evaluating dialysis tubing for dialysis application
- Packaged dry with glycerin (soak in water) or without glycerin (no soaking required)
Specifications
3.5 kDa |