Learn More
Spectrum™ Labs Spectra/Por™ 3 3.5 kD MWCO Standard RC Dry Dialysis Kits
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
Brand:  Spectrum™ 132 720T1
 View more versions of this product
Product Code. 15340772
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Includes:
  • 1m or 5m of Spectra/Por 3 dialysis tubing
  • 1 Standard Spectra/Por Closure (for buoyance)
  • 1 weighted Spectra/Por Closure (for orientation)
  • 2 adhesive tubing openers
Description
Description
  • For basic dialysis applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry with glycerin (soak in water) or without glycerin (no soaking required)
Specifications
Specifications
3.5 kDa
11.5 mm
1.1 mL/cm vol./length
1 m Tubing/Kit