missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Labs Spectra/Por™ 6 10 kD MWCO Standard RC Pre-wetted Dialysis Kits

With Pre-wetted ready-to-use tubing, not requiring soaking to remove glycerol

Brand:  Spectrum™ 132574T

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15390722

  • £110.00 / 1m
Estimated Shipment: 13-08-2024
to see stock.

Includes:
  • 1 meter of Spectra/Por 6 Dialysis Tubing
  • 1 Standard Spectra/Por Closure (for buoyancy)
  • 1 Weighted Spectra/Por Closure (for orientation)
  • 5 Tubing Opening Picks

Description

Description

  • Clear, flexible, study standard grade Regenerated Cellulose (RC) dialysis membrane
  • Membrane packaged wet in preservative solution of 0.05% sodium azide
  • Kits available in 1m tubing lengths ideal for membrane evaluation and qualification
  • Include a pair of closures and convenient tubing opening picks

Specifications

Specifications

10 kDa
20.4 mm
3.3 mL/cm vol./length
1 m Tubing/Kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers