missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs Spectra/Por™ 6 10 kD MWCO Standard RC Pre-wetted Dialysis Kits
With Pre-wetted ready-to-use tubing, not requiring soaking to remove glycerol
£110.00
Specifications
MWCO | 10 kDa |
---|---|
Quantity | 1 m Tubing/Kit |
Includes:
- 1 meter of Spectra/Por 6 Dialysis Tubing
- 1 Standard Spectra/Por Closure (for buoyancy)
- 1 Weighted Spectra/Por Closure (for orientation)
- 5 Tubing Opening Picks
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
15390722
|
Spectrum™
132574T |
3.3 mL/cm vol./length | 20.4 mm |
£110.00
1m |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15380722
|
Spectrum™
132572T |
1.8 mL/cm vol./length | 15 mm |
£110.00
1m |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15300732
|
Spectrum™
132576T |
6.4 mL/cm vol./length | 29 mm |
£110.00
1m |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15370722
|
Spectrum™
132570T |
0.45 mL/cm vol./length | 7.5 mm |
£110.00
1m |
Estimated Shipment: 05-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Clear, flexible, study standard grade Regenerated Cellulose (RC) dialysis membrane
- Membrane packaged wet in preservative solution of 0.05% sodium azide
- Kits available in 1m tubing lengths ideal for membrane evaluation and qualification
- Include a pair of closures and convenient tubing opening picks
Specifications
10 kDa | |
1 m Tubing/Kit |