missing translation for 'onlineSavingsMsg'
Learn More

6-Benzyladenine, 99%, Thermo Scientific Chemicals

£20.20 - £205.02

Chemical Identifiers

CAS 1214-39-7
Molecular Formula C12H11N5
Molecular Weight (g/mol) 225.26
MDL Number MFCD00005572
InChI Key NWBJYWHLCVSVIJ-UHFFFAOYSA-N
Synonym 6-benzylaminopurine, benzyladenine, n6-benzyladenine, n-benzyladenine, n-benzyl-9h-purin-6-amine, 6-benzyladenine, 6-benzylamino purine, cytokinin b, adenine, n-benzyl, 6-bap
PubChem CID 62389
ChEBI CHEBI:29022
IUPAC Name N-benzyl-7H-purin-6-amine
SMILES C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11417666
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14678.03
1 g
£20.20
1g
Please to purchase this item. Need a web account? Register with us today!
11427666
View Documents
Thermo Scientific Alfa Aesar
A14678.06
5 g
£60.60
5g
Please to purchase this item. Need a web account? Register with us today!
11437666
View Documents
Thermo Scientific Alfa Aesar
A14678.14
25 g
£205.02
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables6-Benzyladenine is a plant growth regulator used to stimulate growth in apples, melons, oranges, roses, pears, sweet cherries and white pine trees. It is also used to establish and screen the optimal rapid propagation technology of S. tonkinensis by orthogonal test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Inhibitor of respiratory kinase in plants. Increases post-harvest life of green vegetables6-Benzyladenine is a plant growth regulator used to stimulate growth in apples, melons, oranges, roses, pears, sweet cherries and white pine trees. It is also used to establish and screen the optimal rapid propagation technology of S. tonkinensis by orthogonal test.

Solubility
Soluble in water, methanol and acetone. Slightly soluble in ethyl acetate and dichloromethane and toluene. Insoluble in n-hexane.

Notes
Incompatible with strong oxidizing agent.
Specifications

Chemical Identifiers

1214-39-7
225.26
NWBJYWHLCVSVIJ-UHFFFAOYSA-N
62389
N-benzyl-7H-purin-6-amine
C12H11N5
MFCD00005572
6-benzylaminopurine, benzyladenine, n6-benzyladenine, n-benzyladenine, n-benzyl-9h-purin-6-amine, 6-benzyladenine, 6-benzylamino purine, cytokinin b, adenine, n-benzyl, 6-bap
CHEBI:29022
C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1

Specifications

1214-39-7
99%
MFCD00005572
6-benzylaminopurine, benzyladenine, n6-benzyladenine, n-benzyladenine, n-benzyl-9h-purin-6-amine, 6-benzyladenine, 6-benzylamino purine, cytokinin b, adenine, n-benzyl, 6-bap
NWBJYWHLCVSVIJ-UHFFFAOYSA-N
N-benzyl-7H-purin-6-amine
62389
225.26
6-Benzyladenine
229°C to 233°C
C12H11N5
19406
Soluble in water,methanol and acetone. Slightly soluble in ethyl acetate and dichloromethane and toluene. Insoluble in n-hexane.
C(NC1=C2NC=NC2=NC=N1)C1=CC=CC=C1
225.26
CHEBI:29022
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 214-927-5

missing translation for 'rtecsNumber' : AU6252200

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.