Learn More
6-Chloro-4-methylpyridine-2-carboxylic acid, 95%, Thermo Scientific Chemicals
POA - £262.00
Chemical Identifiers
CAS | 324028-95-7 |
---|---|
Molecular Formula | C7H6ClNO2 |
Molecular Weight (g/mol) | 171.58 |
MDL Number | MFCD09834827 |
InChI Key | IDIRHQNTRMGBPJ-UHFFFAOYSA-N |
Synonym | 6-chloro-4-methyl-2-pyridinecarboxylic acid, 6-chloro-4-methylpicolinic acid, 6-chloro-4-methyl-pyridine-2-carboxylic acid, 6-chloranyl-4-methyl-pyridine-2-carboxylic acid |
PubChem CID | 23002381 |
IUPAC Name | 6-chloro-4-methylpyridine-2-carboxylic acid |
SMILES | CC1=CC(=NC(=C1)Cl)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15478386
|
Thermo Scientific Alfa Aesar
H63584.MD |
250 mg | |||||||
15450349
|
Thermo Scientific Alfa Aesar
H63584.03 |
1 g |
£76.30
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15460349
|
Thermo Scientific Alfa Aesar
H63584.06 |
5 g |
£262.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
324028-95-7 | |
171.58 | |
IDIRHQNTRMGBPJ-UHFFFAOYSA-N | |
23002381 | |
CC1=CC(=NC(=C1)Cl)C(=O)O |
C7H6ClNO2 | |
MFCD09834827 | |
6-chloro-4-methyl-2-pyridinecarboxylic acid, 6-chloro-4-methylpicolinic acid, 6-chloro-4-methyl-pyridine-2-carboxylic acid, 6-chloranyl-4-methyl-pyridine-2-carboxylic acid | |
6-chloro-4-methylpyridine-2-carboxylic acid |
Specifications
324028-95-7 | |
C7H6ClNO2 | |
UN2811 | |
IDIRHQNTRMGBPJ-UHFFFAOYSA-N | |
6-chloro-4-methylpyridine-2-carboxylic acid | |
23002381 | |
95% | |
6-Chloro-4-methylpyridine-2-carboxylic acid |
White | |
MFCD09834827 | |
6-chloro-4-methyl-2-pyridinecarboxylic acid, 6-chloro-4-methylpicolinic acid, 6-chloro-4-methyl-pyridine-2-carboxylic acid, 6-chloranyl-4-methyl-pyridine-2-carboxylic acid | |
CC1=CC(=NC(=C1)Cl)C(=O)O | |
171.58 | |
171.58 | |
Solid |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only