missing translation for 'onlineSavingsMsg'
Learn More
6-Hydroxycaproic acid, 95%, Thermo Scientific Chemicals Product Code.: 13478518

6-Hydroxycaproic acid, 95%, Thermo Scientific Chemicals

Product Code. 13478518
25 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 13478518

Brand: Thermo Scientific Acros 342240250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-25-9
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00046560
InChI Key IWHLYPDWHHPVAA-UHFFFAOYSA-N
Synonym 6-hydroxycaproic acid, 6-hydroxyhexanoate, hexanoic acid, 6-hydroxy, 6-hydroxy caproic acid, 6-hydroxy-hexanoic acid, epsilon-hydroxycaproic acid, 5-carboxypentanol, unii-3y3ox37nm8, 6-hydroxycaproicacid,pract.,containslactone, 6-hydroxy-caproic acidShow MoreShow Less
PubChem CID 14490
ChEBI CHEBI:17869
IUPAC Name 6-hydroxyhexanoic acid
SMILES C(CCC(=O)O)CCO

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 38.0°C to 40.0°C
Color White
Density 0.9810g/mL
Boiling Point 113.0°C to 116.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula OH(CH2)5COOH
Refractive Index 1.44
Quantity 25 g
Specific Gravity 0.981
Formula Weight 132.16
Percent Purity 95%
Physical Form Low Melting Crystalline Solid
Chemical Name or Material 6-Hydroxycaproic acid, 95%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467690 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number342240050,342240250,342240010
Lot NumberA0465679 Certificate TypeCertificate of Analysis Date04/09/2024 Catalog Number342240050,342240250,342240010
Lot NumberA0461636 Certificate TypeCertificate of Analysis Date10/04/2024 Catalog Number342240050,342240250,342240010
Lot NumberA0454080 Certificate TypeCertificate of Analysis Date09/01/2024 Catalog Number342240050,342240250,342240010
Lot NumberA0447331 Certificate TypeCertificate of Analysis Date08/12/2023 Catalog Number342240050,342240250,342240010
compliance-icons
Product Identifier
  • 6-Hydroxycaproic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Hydroxycaproic acid, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.