Learn More
6-Hydroxyhexanoic acid, 95%, may cont. variable amounts of dimer, Thermo Scientific Chemicals
Description
6-Hydroxyhexanoic acid acts as a useful reagent in synthetic chemistry. It is used as a precursor to prepare surface active monomers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
6-Hydroxyhexanoic acid acts as a useful reagent in synthetic chemistry. It is used as a precursor to prepare surface active monomers.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 38°C to 40°C |
Density | 1.08 |
Flash Point | >110°C (230°F) |
Linear Formula | HO(CH2)5CO2H |
Refractive Index | 1.456 |
Quantity | 5 g |
Formula Weight | 132.16 |
Percent Purity | 95% |
Assay | may cont. variable amounts of dimer |
Chemical Name or Material | 6-Hydroxyhexanoic acid |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10220540 | Certificate of Analysis | 30/08/2024 | B24857.06,B24857.03,B24857.14 |
10252073 | Certificate of Analysis | 13/08/2024 | B24857.06,B24857.03,B24857.14 |
10236849 | Certificate of Analysis | 11/12/2023 | B24857.06,B24857.03,B24857.14 |
10220540 | Certificate of Analysis | 23/11/2021 | B24857.06,B24857.03,B24857.14 |
10102927 | Certificate of Analysis | 23/11/2021 | B24857.06,B24857.03,B24857.14 |
Safety and Handling

- 6-Hydroxyhexanoic acid
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : 6-Hydroxyhexanoic acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.